(R)-(-)-1-Phenyl-2-propanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1572-95-8
:
Formula:C₉H₁₂O
:136.19 g/mol
Boiling Pt:219…221 °C
Density:0.973 g/cm³ (25 °C)
Flash Pt:>85 °C
:GHS07
Storage Temperature:Ambient
Product Information
- Catalogus #Leverancier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR150-1GLocal Article Number: APOSOR150-1G
- Supplier Article Number: OR150-5GLocal Article Number: APOSOR150-5G