2-Methyl-2-{[(4-methylphenyl)methyl]amino}propanenitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1018265-76-3
:
Formula:C₁₂H₁₆N₂
MDL Number:MFCD10014475
:188.27 g/mol
Melting Pt:36…40 °C
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR303949-500MGLocal Article Number: APOSOR303949-500MG
- Supplier Article Number: OR303949-1GLocal Article Number: APOSOR303949-1G