Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Netherlands
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
1,3-Dihydropyrrolo[2,3-b]pyridin-2-one
1,3-Dihydropyrrolo[2,3-b]pyridin-2-one
1,3-Dihydropyrrolo[2,3-b]pyridin-2-one
1,3-Dihydropyrrolo[2,3-b]pyridin-2-one
Supplier:
Apollo Scientific
Certificates
Expand image
1,3-Dihydropyrrolo[2,3-b]pyridin-2-one
Supplier:
Apollo Scientific
About this item
CAS Number:
5654-97-7
:
Formula:
C₇H₆N₂O
MDL Number:
MFCD07367951
:
134.14 g/mol
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSOR317324-1G
OR317324-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 24,60 €
24,60 €
-
+
Add to cart
1 * 1 g
24,60 €
-
+
Add to cart
APOSOR317324-5G
OR317324-5G
Bottle
5 g
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 99,40 €
99,40 €
-
+
Add to cart
1 * 5 g
99,40 €
-
+
Add to cart