3-Bromo-6-methoxypyridazine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17321-29-8
:
Formula:C5H5BrN2O
MDL Number:MFCD09702456
:189.0100 g/mol
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR52675-5GLocal Article Number: APOSOR52675-5G
- Supplier Article Number: OR52675-10GLocal Article Number: APOSOR52675-10G