3-(tert-Butyl)-2-hydroxybenzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:24623-65-2
:
Formula:C11H14O2
MDL Number:MFCD00778877
:178.2277 g/mol
Boiling Pt:78…79 °C/1mmHg
Density:1.041 g/cm³ (25 °C)
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR6969-1GLocal Article Number: APOSOR6969-1G
- Supplier Article Number: OR6969-5GLocal Article Number: APOSOR6969-5G
- Supplier Article Number: OR6969-25GLocal Article Number: APOSOR6969-25G