2-Methylhippuric acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:42013-20-7
:
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD00050991
:193.1992 g/mol
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR921087-1GLocal Article Number: APOSOR921087-1G
- Supplier Article Number: OR921087-5GLocal Article Number: APOSOR921087-5G
- Supplier Article Number: OR921087-25GLocal Article Number: APOSOR921087-25G