Netherlands
Order Entry
6,7-Dihydro-5H-cyclopenta[b]pyridin-3-amine 98%
6,7-Dihydro-5H-cyclopenta[b]pyridin-3-amine 98%
Supplier:  Apollo Scientific
Certificates
6,7-Dihydro-5H-cyclopenta[b]pyridin-3-amine 98%
Supplier:  Apollo Scientific
Product Information
  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • APOSOR922328-250MG
    OR922328-250MG
    Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 387,00 €
    387,00 €
    1 * 250 mg
       387,00 €
    Supplier Article Number: OR922328-250MG
    Local Article Number: APOSOR922328-250MG
  • APOSOR922328-1G
    OR922328-1G
    Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 992,00 €
    992,00 €
    1 * 1 g
       992,00 €
    Supplier Article Number: OR922328-1G
    Local Article Number: APOSOR922328-1G