Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Netherlands
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
1-Methoxy-1-trimethylsilyloxypropene
1-Methoxy-1-trimethylsilyloxypropene 95%
1-Methoxy-1-trimethylsilyloxypropene 95%
1-Methoxy-1-trimethylsilyloxypropene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1-Methoxy-1-trimethylsilyloxypropene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
34880-70-1
:
Formula:
C₇H₁₆O₂Si
MDL Number:
MFCD00671554
:
160.29 g/mol
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSOR924980-250MG
OR924980-250MG
Bottle
250 mg
Log in
for availability
Log in
for availability
1 * 250 mg
1 * 250 mg - 169,00 €
169,00 €
-
+
Add to cart
1 * 250 mg
169,00 €
-
+
Add to cart
APOSOR924980-1G
OR924980-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 431,00 €
431,00 €
-
+
Add to cart
1 * 1 g
431,00 €
-
+
Add to cart