Netherlands
Order Entry
3-(5-Amino-3-methyl-1H-pyrazol-1-yl)propanenitrile 95%
3-(5-Amino-3-methyl-1H-pyrazol-1-yl)propanenitrile 95%
Supplier:  Apollo Scientific
Certificates
3-(5-Amino-3-methyl-1H-pyrazol-1-yl)propanenitrile 95%
Supplier:  Apollo Scientific
Product Information
  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • APOSOR951441-250MG
    OR951441-250MG
    Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 191,00 €
    191,00 €
    1 * 250 mg
       191,00 €
    Supplier Article Number: OR951441-250MG
    Local Article Number: APOSOR951441-250MG
  • APOSOR951441-1G
    OR951441-1G
    Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 438,00 €
    438,00 €
    1 * 1 g
       438,00 €
    Supplier Article Number: OR951441-1G
    Local Article Number: APOSOR951441-1G