2-Chloro-1-fluoro-4-nitrobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:350-30-1
:
Formula:C₆H₃ClFNO₂
MDL Number:MFCD00007206
:175.55 g/mol
Boiling Pt:227…232 °C
Melting Pt:40…45 °C
Density:1.61 g/cm³ (25 °C)
Flash Pt:>110 °C
EINECS:206-499-3
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: PC1870-100GLocal Article Number: APOSPC1870-100G
- Supplier Article Number: PC1870-500GLocal Article Number: APOSPC1870-500G
- Supplier Article Number: PC1870-1KGLocal Article Number: APOSPC1870-1KG