Netherlands
Order Entry
3-Bromo-2,6-dichloro-5-(trifluoromethyl)phenylacetic acid
3-Bromo-2,6-dichloro-5-(trifluoromethyl)phenylacetic acid
Supplier:  Apollo Scientific
Certificates
3-Bromo-2,6-dichloro-5-(trifluoromethyl)phenylacetic acid
Supplier:  Apollo Scientific
Product Information
  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • APOSPC303502-1G
    PC303502-1G
    Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 421,00 €
    421,00 €
    1 * 1 g
       421,00 €
    Supplier Article Number: PC303502-1G
    Local Article Number: APOSPC303502-1G
  • APOSPC303502-5G
    PC303502-5G
    Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 982,00 €
    982,00 €
    1 * 5 g
       982,00 €
    Supplier Article Number: PC303502-5G
    Local Article Number: APOSPC303502-5G