Netherlands
Order Entry
J.T.Baker® BAKER INSTRA-ANALYZED®, Gold Plasma Standards
 :  Avantor
J.T.Baker® BAKER INSTRA-ANALYZED®, Gold Plasma Standards
Supplier:  Avantor
Product Information
  • Catalog #
    Element
    Symbol
    Concentration
    Matrix
    Pk
    Availability
    Price
  • 5763.0100
    Gold
    Au
    1000 mg/L
    5% HCl
    100 mL
    Log in for availability
    1 * 100 mL
    1 * 100 mL
     : BAKR5763.0100
  • 5730.0100
    Gold
    Au
    10000 mg/L
    5% HCl
    100 mL
    1 * 100 mL
    1 * 100 mL
     : BAKR5730.0100