J.T.Baker® Plate 96 PCR
Leverancier: Ritter, part of Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43001-0300EA 159.2 EUR
BAKR43001-0300
J.T.Baker® Plate 96 PCR
White plate for real-time PCR.
  • Thin-walled wells provide a good heat transfer
  • Working volume of max. 200 µl
  • Raised rims to minimize the risk of cross-contamination
  • Ideal for the use with self-adhesive or heat-sealing foils
  • Alphanumeric code for easy identification of certain samples
J.T.Baker® Plate 96 PCR is designed for the use with Roche LightCycler 480.