dNTP set
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
APOSBIA2593-4X0.25EA 0 EUR
APOSBIA2593-4X0.25
dNTP set
Reagentia voor nucleïnezuren Nucleotiden
Four Separate solutions of the sodium salts of the deoxyribonucleotide triphosphates (dATP, dCTP, dGTP, dTTP), 100 mM each, phosphate buffered pH 7,5.