Supelco® Ascentis® Express PFAS HPLC Columns
Supplier: Merck
Certificates
About this item
Ascentis® Express PFAS HPLC columns, with Fused-Core® technology delivers necessary retention to perform in EPA methods 537.1, 533 and 8327. Fast and high resolution LC-MS separation of PFAS.
- 2,7 µm Fused-Core® particle for reliable and high efficiency separations and lower column back pressure compared to sub 2 particles
- Excellent suitability for MS detection
- Application-related lot analysis and single column performance testing
The U.S. Food and Drug Administration (FDA) and U.S. Environmental Protection Agency (EPA) have initiated actions against PFAS. For determination of PFASs, liquid chromatography-mass spectrometry (LC-MS) is a commonly used technique.
Delivery information: Supplied with column test certificate inside the box.
Caution: The delay column is used with a larger ID than the analytical column:
- Delay column 3,0 mm int. Ø is used with Analytical column 2,1 mm int. Ø
- Delay column 4,6 mm int. Ø is used with Analytical column 3 mm int. Ø
- Delay column 3,0 mm int. Ø is used with Analytical column 2,1 mm int. Ø
- Delay column 4,6 mm int. Ø is used with Analytical column 3 mm int. Ø
Specifications
- Catalog No:
- 53570-U
- 53565-U
- 53573-U
- 53572-U
- 53557-U
- 53559-U
- 53564-U
- 53562-U
- 53560-U
- 53563-U
- Application:
- PFAS separation
- PFAS separation
- PFAS separation
- PFAS separation
- PFAS separation
- PFAS separation
- PFAS separation
- PFAS separation
- PFAS separation
- PFAS separation
- Carbon load:
- 9,9%
- 9,9%
- 9,9%
- 9,9%
- 9,9%
- 9,9%
- 9,9%
- 9,9%
- 9,9%
- 9,9%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Surface area:
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- 135 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
Specifications
2,7 µm HPLC columns
Product Information
- Catalog #Supplier #Column FormatPhaseColumn hardwarePore sizeParticle SizeØ int.AvailabilityPrice
- 53565-U53565-UAnalyticalC18 (Octadecyl)Stainless steel90 Å2,7 µm3,0 mmLog in for availability1 * 1 ST796,00 €Supplier Article Number: 53565-ULocal Article Number: SUPL53565-USpecifications:
More
Accessories
Specifications
Environmentally PreferableApplicationPFAS separationCarbon load9,9%EndcappedYesMax. pressure600 barpH range2 - 9Surface area135 m²/gUSP codeL1 - 53570-U53570-UAnalyticalC18 (Octadecyl)Stainless steel90 Å2,7 µm3,0 mmLog in for availability1 * 1 ST1 010,00 €Supplier Article Number: 53570-ULocal Article Number: SUPL53570-USpecifications:
More
Accessories
Specifications
Environmentally PreferableApplicationPFAS separationCarbon load9,9%EndcappedYesMax. pressure600 barpH range2 - 9Surface area135 m²/gUSP codeL1 - 53557-U53557-UAnalyticalC18 (Octadecyl)Stainless steel90 Å2,7 µm2,1 mmLog in for availability1 * 1 ST614,00 €Supplier Article Number: 53557-ULocal Article Number: SUPL53557-USpecifications:
More
Accessories
Specifications
Environmentally PreferableApplicationPFAS separationCarbon load9,9%EndcappedYesMax. pressure600 barpH range2 - 9Surface area135 m²/gUSP codeL1 - 53559-U53559-UAnalyticalC18 (Octadecyl)Stainless steel90 Å2,7 µm2,1 mmLog in for availability1 * 1 ST838,00 €Supplier Article Number: 53559-ULocal Article Number: SUPL53559-USpecifications:
More
Accessories
Specifications
Environmentally PreferableApplicationPFAS separationCarbon load9,9%EndcappedYesMax. pressure600 barpH range2 - 9Surface area135 m²/gUSP codeL1 - 53560-U53560-UAnalyticalC18 (Octadecyl)Stainless steel90 Å2,7 µm2,1 mmLog in for availability1 * 1 ST924,00 €Supplier Article Number: 53560-ULocal Article Number: SUPL53560-USpecifications:
More
Accessories
Specifications
Environmentally PreferableApplicationPFAS separationCarbon load9,9%EndcappedYesMax. pressure600 barpH range2 - 9Surface area135 m²/gUSP codeL1 - 53562-U53562-UAnalyticalC18 (Octadecyl)Stainless steel90 Å2,7 µm2,1 mmLog in for availability1 * 1 ST1 010,00 €Supplier Article Number: 53562-ULocal Article Number: SUPL53562-USpecifications:
More
Accessories
Specifications
Environmentally PreferableApplicationPFAS separationCarbon load9,9%EndcappedYesMax. pressure600 barpH range2 - 9Surface area135 m²/gUSP codeL1 - 53563-U53563-UAnalyticalC18 (Octadecyl)Stainless steel90 Å2,7 µm3,0 mmLog in for availability1 * 1 ST563,00 €Supplier Article Number: 53563-ULocal Article Number: SUPL53563-USpecifications:
More
Accessories
Specifications
Environmentally PreferableApplicationPFAS separationCarbon load9,9%EndcappedYesMax. pressure600 barpH range2 - 9Surface area135 m²/gUSP codeL1 - 53564-U53564-UAnalyticalC18 (Octadecyl)Stainless steel90 Å2,7 µm3,0 mmLog in for availability1 * 1 ST838,00 €Supplier Article Number: 53564-ULocal Article Number: SUPL53564-USpecifications:
More
Accessories
Specifications
Environmentally PreferableApplicationPFAS separationCarbon load9,9%EndcappedYesMax. pressure600 barpH range2 - 9Surface area135 m²/gUSP codeL1
2,7 µm Delay HPLC columns
Product Information
- Catalog #Supplier #Column FormatPhaseColumn hardwarePore sizeParticle SizeØ int.AvailabilityPrice
- Supplier Article Number: 53572-ULocal Article Number: SUPL53572-USpecifications:
More
Accessories
Specifications
Environmentally PreferableApplicationPFAS separationCarbon load9,9%EndcappedYesMax. pressure600 barpH range2 - 9Surface area135 m²/gUSP codeL1 - Supplier Article Number: 53573-ULocal Article Number: SUPL53573-USpecifications:
More
Accessories
Specifications
Environmentally PreferableApplicationPFAS separationCarbon load9,9%EndcappedYesMax. pressure600 barpH range2 - 9Surface area135 m²/gUSP codeL1
Guard cartridge holder
Product Information
- Catalog #DescriptionPkAvailabilityPrice
- Guard cartridge holder, for use with Ascentis® Express guard columns (guard cartridge not included with holder)1Log in for availabilitySupplier Article Number: 53500-ULocal Article Number: SUPL53500-U