Transport and storage containers, multi-use box, VWR®
Leverancier: VWR, part of Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
216-2383EA 68.3 EUR
216-2383 216-2382 216-2381 216-2379 216-2378 216-2380
Transport and storage containers, multi-use box, VWR®
Dozen Transportdozen
Made of transparent virgin polypropylene which is food grade. Multi-use boxes are strong, transparent plastic containers that can be used to pack, store and transport a wide variety of products, including foodstuffs.
  • Transparent colour makes the contents easily visible
  • Hygienic smooth walls for easy cleaning
  • Food grade, freezer and dishwasher safe
  • Suitable for –25 to +95 °C storage
  • Stackable when lid is in place, nestable without the lids when not in use
  • 90% space saving when empty
  • Lid is secured in place with the clip handles
Available in large, medium and small sizes to suit all your requirements. Their ergonomic clip handles allow for a comfortable and secure grip during storage and transport, and their lids offer a perfect stability when stacking.