U heeft gezocht op: Regelaars
17  results were found
SearchResultCount:"17"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: SI Analytics
Omschrijving: The temperature of capillary viscometers can be stabilised in this jacket using circulation thermostats from 0 to 180 °C.
Leverancier: JULABO GmbH
Omschrijving: Temperatuurafhankelijke meet-, beheers-, veiligheids- en controletaken uitvoeren. Typische toepassingen: verwarmingsmantels, verwarmingsbaden en verwa...
Leverancier: Cole-Parmer
Omschrijving: Cost-effective control of resistive heating loads.
Catalogus nummer: (WINKWRT00011-XX010)
Leverancier: WINKLER GMBH
Omschrijving: Plug-in power selector made from polystyrene with safety socket with integrated fuse.
UOM: 1 * 1 ST
Leverancier: Heidolph Instruments GmbH & Co.KG
Omschrijving: Electronic temperature controller with innovative design, can be combined with Hei-Standard (old models), and MR 3001 K and MR 3002 models. The temper...
Catalogus nummer: (HUBE9771)
Leverancier: HUBER
Omschrijving: This breather controller enables small pressurisation of the expansion vessel.
UOM: 1 * 1 ST
Leverancier: Industrial Physics
Omschrijving: The temperature control jacket for viscosity cups is a double jacketed casing of anodised aluminium with built-in spirit level to bring the viscosity ...
Product and/or packaging contains recycled/renewable content
Catalogus nummer: (WINKWRT00116-XX013)
Leverancier: WINKLER GMBH
Omschrijving: Plug-in power selector made of PS with switching mechanism.
UOM: 1 * 1 ST
Catalogus nummer: (STUA04807-90)
Leverancier: Cole Parmer
Omschrijving: All hotplate models are compatible with the Stuart temperature controller SCT2. The SCT2 provides precise temperature control in aqueous or oily sampl...
UOM: 1 * 1 ST
Catalogus nummer: (HUBE10503)
Leverancier: HUBER
Omschrijving: This gateway enables the connection of temperature control units to Profibus systems.
UOM: 1 * 1 ST
Catalogus nummer: (WINKWRT00117-XX010)
Leverancier: WINKLER GMBH
Omschrijving: Plug-in power selector made of polystyrene with switching mechanism.
UOM: 1 * 1 ST
Catalogus nummer: (ELET32587-00)
Leverancier: Cole-Parmer
Omschrijving: Get accurate temperature control of heating equipment in your lab.
UOM: 1 * 1 ST
Catalogus nummer: (453-2108)
Leverancier: GESTIGKEIT HARRY
Omschrijving: TR 28-3T is a regulating unit for high-temperature hotplates.
UOM: 1 * 1 ST
Leverancier: Carbolite
Omschrijving: These controllers for programmable temperature can use up to eight segments, each segment comprising a ramp followed by a dwell. The dwell may be set ...
Catalogus nummer: (453-2109)
Leverancier: GESTIGKEIT HARRY
Omschrijving: The micro-processor controlled ramp controller is a complex time/temperature control and this program is indefinitely stored by means of an EEPROM.
UOM: 1 * 1 ST
Catalogus nummer: (VELPF208B0063)
Leverancier: VELP SCIENTIFIC
Omschrijving: Digital thermoregulator used to combine VTF with PW 10 derivation non VELP devices. Unit has fuzzy logic technology and includes two clamps for height...
UOM: 1 * 1 ST
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 17
no targeter for Bottom