Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Criteria
    • In Stock (4)
  • Leverancier
  • Refine by Suppliers
    Sort by:
  • Zoek binnen resultaten
Borden voor dissectie
https://nl.vwr.com/stibo/smallweb/std.lang.all/51/33/18945133.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"6"
  List view Grid view Easy View (new)
Sorteer op:
Leverancier:  Simport Scientific
Omschrijving:   Plankje met drie snijgebieden, DissecTable™, PE, wit, b×d×h: 330,2×279,4×12,5 mm
Leverancier:  Simport Scientific
Omschrijving:   These dissection boards are made from a copolymer resin in order to increase impact and temperature performance. The board is strong yet inexpensive e...
Catalogus nummer: (216-1300)
Leverancier:  SCILABWARE
Omschrijving:   Durable cutting board made of PP.
UOM:  1 * 1 ST
Leverancier:  LP ITALIANA SPA
Omschrijving:   Disposable cutting boards made of cardboard and PS foam for the dissection of surgical specimens.
Leverancier:  MURRAY, S & CO LTD
Omschrijving:   Wooden dissecting boards with frame edge.
Leverancier:  SEMADENI
Omschrijving:   These chopping boards are made from professional-grade HYGENIA® HDPE 500 material. Ideal for the food-service and food-processing industries.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 6  Of 6
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping