U heeft gezocht op: Valbescherming
88  results were found
SearchResultCount:"88"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (MMMA3504426)
Leverancier: 3M
Omschrijving: Anchorage connectors are built from top quality materials guaranteeing high strength and durability to stand up to the toughest environments. Anchorag...
UOM: 1 * 1 ST
New Product
Leverancier: 3M
Omschrijving: The 3M™ DBI SALA® Clip2Loop Coil Tool Tether is engineered to withstand harsh work environments.
New Product
Leverancier: 3M
Omschrijving: Cross chest buckle assembly for central front fall arrest attachment point - optimised position on body. Revolving torso adjusters (patented) for quic...
New Product
Leverancier: 3M
Omschrijving: 3M DBI-SALA ExoFit XE200 harness with rescue loops, automatic buckles, two sizes. X-shaped chest straps with central D-ring attachment point optimised...
New Product
Leverancier: CAPITAL SAFETY DEUTSCHLAND
Omschrijving: Protecta® work positioning rope connecting lanyards are an ideal solution for the connecting element in a restraint system. Made from polyamide braide...
Catalogus nummer: (CPSD1500068)
Leverancier: CAPITAL SAFETY DEUTSCHLAND
Omschrijving: Lightweight vinyl construction - ideal alternative to classic bungee tethers.
UOM: 1 * 10 ST
Catalogus nummer: (PRTWFP14RER)
Leverancier: Portwest
Omschrijving: Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collisio...
UOM: 1 * 1 ST
Leverancier: CAPITAL SAFETY DEUTSCHLAND
Omschrijving: The Protecta® Comfort Belt Style Fall Arrest Harness is designed to provide reliable protection, comfort, and convenience. It now features upgraded fe...
Leverancier: CAPITAL SAFETY DEUTSCHLAND
Omschrijving: Protecta® Standard Vest Style Fall Arrest Harness is a sleek and modern safety harness with upgraded features. It has water and stain resistant webbin...
Leverancier: CAPITAL SAFETY DEUTSCHLAND
Omschrijving: Sling polyester webbing and stainless steel links.
Catalogus nummer: (CPSDAM211)
Leverancier: CAPITAL SAFETY DEUTSCHLAND
Omschrijving: Protecta® anchorage D-ring made from 316 stainless steel; 12 mm fixing.
UOM: 1 * 1 ST
Leverancier: CAPITAL SAFETY DEUTSCHLAND
Omschrijving: The Rollgliss R350 system provides efficient hauling, but more importantly, very controlled lowering (a finger and thumb are all that is required to h...
Catalogus nummer: (CPSD1150492)
Leverancier: CAPITAL SAFETY DEUTSCHLAND
Omschrijving: PROTECTA® Gear loops are a part of the fall arrest harness accessory options. They provide the opportunity to add extra tool carrying convenience to a...
UOM: 1 * 2 ST
Catalogus nummer: (CPSD1500009)
Leverancier: CAPITAL SAFETY DEUTSCHLAND
Omschrijving: D-Ring cord attachment point, 2,3 kg (5 lbs.) capacity.
UOM: 1 * 10 ST
Leverancier: CAPITAL SAFETY DEUTSCHLAND
Omschrijving: This pole hoist attaches to a carabiner on the universal bracket to a suitable anchor point, allowing to work in any direction from any angle.
Catalogus nummer: (CPSD8000118)
Leverancier: CAPITAL SAFETY DEUTSCHLAND
Omschrijving: The DBI-SALA® Confined space HC (high capacity) short reach davit system is constructed of powder-coated aluminium, and features a modular design for ...
UOM: 1 * 1 EA
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 88
no targeter for Bottom