Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Criteria
  • Leverancier
  • Refine by Suppliers
    Sort by:
  • Zoek binnen resultaten
Persmachines
https://nl.vwr.com/stibo/smallweb/std.lang.all/96/77/18989677.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"6"
  List view Easy View (new)
Sorteer op:
Catalogus nummer: (43661.KT)
Leverancier:  Alfa Aesar
Omschrijving:   Macro/micro KBr pellet die set, 13 mm.
UOM:  1 * 1 ST
Catalogus nummer: (RETS20.750.0006)
Leverancier:  Retsch GmbH
Omschrijving:   The PP 25 is a benchtop unit with simple and safe operation.
UOM:  1 * 1 ST
Leverancier:  Retsch GmbH
Omschrijving:   High-quality pellets are a precondition for X-ray fluorescence (XRF) analysis.
Catalogus nummer: (PERC51001144)
Leverancier:  PERKIN ELMER GC-INSTRUMENTS
Omschrijving:   The evacuable potassium bromide (KBr) die is designed to produce uniform KBr pellets.
UOM:  1 * 1 ST
Catalogus nummer: (PERC51004599)
Leverancier:  PERKIN ELMER GC-INSTRUMENTS
Omschrijving:   This compact hydraulic press is used with the KBr die to prepare KBr pellets.
UOM:  1 * 1 ST
Catalogus nummer: (SHIM200-66747-91)
Leverancier:  SHIMADZU
Omschrijving:   This compact mini hand press is a used to produce KBr pellets.
UOM:  1 * 1 ST
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 6  Of 6