U heeft gezocht op: Kleurstoffen
160  results were found
SearchResultCount:"160"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Merck
Omschrijving: Entellan® New is een nieuw montagemedium voor dekglaasjes en is vooral geschikt voor commerciële automatische inbouwinstrumenten die werken met dekgla...
Leverancier: SIGMA-ALDRICH MICROSCOPY
Omschrijving: Microscopy and staining reagents, Immersion oil 150 cSt
Catalogus nummer: (317616-100ML)
Leverancier: SIGMA-ALDRICH MICROSCOPY
Omschrijving: Microscopy and staining reagents, Neutral mounting medium, DPX
UOM: 1 * 100 mL
Leverancier: SIGMA-ALDRICH MICROSCOPY
Omschrijving: Immersion oil is used for high resolution (1000X) light microscopy work under oil immersion objective lens.
Catalogus nummer: (1.00579.0500)
Leverancier: Merck
Omschrijving: DPX new is a synthetic non aqueous mounting medium for microscopy. In its formulation, the teratogenic plasticiizer dibutyl phthalate (DBP) has been a...
UOM: 1 * 500 mL
MSDS
Leverancier: Merck
Omschrijving: Harde weefsels, zoals botten of tanden en alle monsters van verkalkt weefsel zoals beenmergbiopsieën, vereisen een speciale voorbehandeling voordat ro...
MSDS Sale
Catalogus nummer: (F4680-25ML)
Leverancier: SIGMA-ALDRICH MICROSCOPY
Omschrijving: Fluoromountâ„¢ Aqueous Mounting Medium for use with fluorescent dye-stained tissue.
UOM: 1 * 25 mL
Catalogus nummer: (F6937-20ML)
Leverancier: SIGMA-ALDRICH MICROSCOPY
Omschrijving: Fluoroshieldâ„¢ with 1,4-Diazabicyclo[2.2.2]octane is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
UOM: 1 * 20 mL
Catalogus nummer: (P0091-6X120ML)
Leverancier: SIGMA-ALDRICH MICROSCOPY
Omschrijving: PolyFreeze is a water-soluble support matrix and a form of embedding medium for frozen sectioning. PolyFreeze sections flawlessly at routine frozen sp...
UOM: 1 * 720 mL
Leverancier: SIGMA-ALDRICH MICROSCOPY
Omschrijving: Immersion oil has been used in epifluorescent microscopy and cell identification.
Leverancier: Merck
Omschrijving: Used to increase lens effectiveness
MSDS Sale
Leverancier: SIGMA-ALDRICH MICROSCOPY
Omschrijving: Araldite® is an epoxy resin commonly used for embedding. It is basically free from polymerization damage and the plastic does not sublime in the micro...
Leverancier: SIGMA-ALDRICH MICROSCOPY
Omschrijving: This is a mounting medium (contains 45% acrylic resin and 55% xylenes) used in microscopy. It is an adhesive and specimen preservative that can be use...
Leverancier: SIGMA-ALDRICH MICROSCOPY
Omschrijving: to be used with SIAL 78711 RenLam M-1 (Epoxy Embedding Medium)
Leverancier: SIGMA-ALDRICH MICROSCOPY
Omschrijving: Glycerol alters the hygroscopic property of gelatin and its permeability to water vapour. Glycerol gelatin or glycerine jelly medium is an aqueous sli...
Leverancier: Merck
Omschrijving: Watervrije montagemedia, zoals Neo-Mount®, moeten worden gebruikt voor permanente montage en langdurige opslag van monsters. Objectglaasjes die in pat...
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 160
no targeter for Bottom