Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Criteria
  • Leverancier
  • Refine by Suppliers
    Sort by:
  • Zoek binnen resultaten
Spuitpompen
https://nl.vwr.com/stibo/smallweb/std.lang.all/69/50/20236950.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"8"
  List view Grid view Easy View (new)
Sorteer op:
Catalogus nummer: (612-5484)
Leverancier:  B. Braun Melsungen AG
Omschrijving:   B. Braun's Perfusor® Space Infusion Pump — with advanced delivery features that offer safety and accuracy — is ideal for operating rooms, adult care, ...
UOM:  1 * 1 ST
Catalogus nummer: (612-5485)
Leverancier:  B. Braun Melsungen AG
Omschrijving:   The volumetric infusion pump Infusomat® Space is one of the basic building blocks of the Space system, designed for acute care adult and pediatric fac...
UOM:  1 * 1 ST
Leverancier:  ISMATEC PART OF COLE PARMER
Omschrijving:   The KDS syringe pumps give customers a cost effective solution for infusing fluids
Catalogus nummer: (ISMCKDS210)
Leverancier:  ISMATEC PART OF COLE PARMER
Omschrijving:   The KDS 210 including five operating modes plus independent rate and volume settings for both infusion and withdrawal.
UOM:  1 * 1 ST
Catalogus nummer: (ISMCKDLEGATO-100)
Leverancier:  ISMATEC PART OF COLE PARMER
Omschrijving:   Infuse only single syringe pump which accommodates syringes from 0,5 µl to 60 ml.
UOM:  1 * 1 ST
Catalogus nummer: (ISMCKDLEGATO-200)
Leverancier:  ISMATEC PART OF COLE PARMER
Omschrijving:   It is an infiuse only dual syringe pump and can accomodate two syringes.
UOM:  1 * 1 ST
Leverancier:  HARVARD APPARATUS LTD
Omschrijving:   The PHD 22/2000 syringe pump series gives you the lowest flow rates, the highest accuracy, the smoothest flow, advanced programmability from the keypa...
Leverancier:  HARVARD APPARATUS LTD
Omschrijving:   This OEM series of syringe pumps have a new mechanism that includes a tight gripping, more secure syringe clamp for syringes ranging from 0,5 µl to 10...
UOM:  1 * 1 ST
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8  Of 8