PCR expansion
Leverancier: Oxford Nanopore Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OXNTEXP-PCA001EA 431 EUR
OXNTEXP-PCA001
PCR expansion
Reagentia voor nucleïnezuren Next generation reagentia voor sequencing
PCR Primers (PRM) and PCR Adapters (PCA).
  • Additional PCR primers and adapters for use with Ligation Sequencing Kits
This kit contains PCR Primers and PCR Adapters, sufficient for at least 24 reactions, and is compatible for use with the Ligation Sequencing Kits (OXNTSQK-LSK110 and OXNTSQK-LSK109).
Leveringsinformatie: Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
Opgelet: Oxford Nanopore Technologies products are currently for research use only.
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.