Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 65-mm, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Don’t sacrifice suitability for precision.
- Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX03216-88
- MFLX03218-41
- MFLX03293-40
- MFLX03218-39
- MFLX03216-36
- MFLX03216-90
- MFLX03293-38
- MFLX03216-38
- MFLX03218-37
- MFLX03216-40
- Model:
- 03216-88
- 03218-41
- 03293-40
- 03218-39
- 03216-36
- 03216-90
- 03293-38
- 03216-38
- 03218-37
- 03216-40
- Accuracy:
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- Float material:
- Carboloy
- Carboloy
- 316 SS
- Glass
- Sapphire
- 316 SS
- 316 SS
- Glass
- Carboloy
- Flow Tube material:
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Max. operating temperature:
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- O-Ring material:
- PTFE
- Viton®
- Buna-N
- Viton®
- Buna-N
- PTFE
- Buna-N
- Buna-N
- Viton®
- Buna-N
- Repeatability:
- ±0,25% full-scale
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Specifications
Product Information
- Catalog #Supplier #Maximum Flow RateFluid typeValve materialFitting materialHousing materialProcess connectionAvailabilityPrice
- 65-mm Correlated Flowmeter, w/ valve, aluminum, 23,169 L/min air65-mm Correlated Flowmeter, w/ valve, aluminum, 23,169 L/min air-23,169 L/minAirAluminumAluminumAluminium0,125" NPT(F)Log in for availability1 * 1 ST180,00 €Specifications:
More
Accessories
Specifications
Model03216-36Environmentally PreferableAccuracy±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glassMax. operating temperature121 °C (250 °F)O-Ring materialBuna-NRepeatability±0,25%Warranty1 year - 65-mm Correlated Flowmeter, w/ valve, aluminum, 42,094 L/min air65-mm Correlated Flowmeter, w/ valve, aluminum, 42,094 L/min air-42,094 L/minAirAluminumAluminumAluminium0,125" NPT(F)Log in for availability1 * 1 ST180,00 €Specifications:
More
Accessories
Specifications
Model03216-38Environmentally PreferableAccuracy±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glassMax. operating temperature121 °C (250 °F)O-Ring materialBuna-NRepeatability±0,25%Warranty1 year - 65-mm Correlated Flowmeter, w/ valve, aluminum, 58,5 L/min air65-mm Correlated Flowmeter, w/ valve, aluminum, 58,5 L/min air03216-4058,5 L/minAirAluminumAluminumAluminium0,125" NPT(F)Log in for availability1 * 1 ST180,00 €Specifications:
More
Accessories
Specifications
Model03216-40Environmentally PreferableAccuracy±2% full-scaleFloat materialCarboloyFlow Tube materialBorosilicate glassMax. operating temperature121 °C (250 °F)O-Ring materialBuna-NRepeatability±0,25%Warranty1 year - 65mm, Correlated Glass/PTFE Flowmeter, with valve, 522 ml/min water65mm, Correlated Glass/PTFE Flowmeter, with valve, 522 ml/min water03216-880,522 L/minWaterPTFE and Kel-F®PTFE and Kel-F®PTFE0,125" NPT(F)Log in for availability1 * 1 ST388,00 €Specifications:
More
Accessories
Specifications
Model03216-88Environmentally PreferableAccuracy±2% full-scaleFloat materialFlow Tube materialBorosilicate glassMax. operating temperature65 °C (150 °F)O-Ring materialPTFERepeatability±0,25% full-scaleWarranty1 year - 65mm, Correlated Glass/PTFE Flowmeter, with valve, 798 ml/min water65mm, Correlated Glass/PTFE Flowmeter, with valve, 798 ml/min water-0,798 L/minWaterPTFE and Kel-F®PTFE and Kel-F®PTFE0,125" NPT(F)Log in for availability1 * 1 ST388,00 €Specifications:
More
Accessories
Specifications
Model03216-90Environmentally PreferableAccuracy±2% full-scaleFloat materialSapphireFlow Tube materialBorosilicate glassMax. operating temperature65 °C (150 °F)O-Ring materialPTFERepeatability±0,25%Warranty1 year - 65-mm Correlated Flowmeter, w/ valve, 316SS, 23,169 L/min air65-mm Correlated Flowmeter, w/ valve, 316SS, 23,169 L/min air-23,169 L/minAir316 SS316 stainless steel316 stainless steel0,125" NPT(F)Log in for availability1 * 1 ST348,00 €Specifications:
More
Accessories
Specifications
Model03218-37Environmentally PreferableAccuracy±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glassMax. operating temperature121 °C (250 °F)O-Ring materialViton®Repeatability±0,25%Warranty1 year - 65-mm Correlated Flowmeter, w/ valve, 316SS, 42,094 L/min air65-mm Correlated Flowmeter, w/ valve, 316SS, 42,094 L/min air03218-3942,094 L/minAir316 SS316 stainless steel316 stainless steel0,125" NPT(F)Log in for availability1 * 1 ST348,00 €Specifications:
More
Accessories
Specifications
Model03218-39Environmentally PreferableAccuracy±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glassMax. operating temperature121 °C (250 °F)O-Ring materialViton®Repeatability±0,25%Warranty1 year - 65-mm Correlated Flowmeter, w/ valve, 316SS, 58,5 L/min air65-mm Correlated Flowmeter, w/ valve, 316SS, 58,5 L/min air-58,5 L/minAir316 SS316 stainless steel316 stainless steel0,125" NPT(F)Log in for availability1 * 1 ST348,00 €Specifications:
More
Accessories
Specifications
Model03218-41Environmentally PreferableAccuracy±2% full-scaleFloat materialCarboloyFlow Tube materialBorosilicate glassMax. operating temperature121 °C (250 °F)O-Ring materialViton®Repeatability±0,25%Warranty1 year - 65-mm Correlated Flowmeter, w/ valve, brass, 42,094 L/min air65-mm Correlated Flowmeter, w/ valve, brass, 42,094 L/min air03293-3842,094 L/minAirChrome-plated brassChrome-plated brassBrass0,125" NPT(F)Log in for availability1 * 1 ST318,00 €Specifications:
More
Accessories
Specifications
Model03293-38Environmentally PreferableAccuracy±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glassMax. operating temperature121 °C (250 °F)O-Ring materialBuna-NRepeatability±0,25%Warranty1 year - 65-mm Correlated Flowmeter, w/ valve, brass, 58,5 L/min air65-mm Correlated Flowmeter, w/ valve, brass, 58,5 L/min air03293-4058,5 L/minAirChrome-plated brassChrome-plated brassBrass0,125" NPT(F)Log in for availability1 * 1 ST318,00 €Specifications:
More
Accessories
Specifications
Model03293-40Environmentally PreferableAccuracy±2% full-scaleFloat materialCarboloyFlow Tube materialBorosilicate glassMax. operating temperature121 °C (250 °F)O-Ring materialBuna-NRepeatability±0,25%Warranty1 year