Netherlands
Order Entry
Flux Pumps Sanitary Drum Pump Air Motors
Flux Pumps Sanitary Drum Pump Air Motors
Certificates
undefined
Flux Pumps Sanitary Drum Pump Air Motors
Specifications
  • Air consumption:
  • 28,9 m³/h (17 ft.³/min)
  • Max. viscosity:
  • 1200 cP
  • 1200 cP
  • Motor hp:
  • 1/2
  • 1/2
  • Warranty:
  • 6 Months
  • 6 Months
Specifications
Product Information
  • Catalogus #
    Leverancier #
    Power
    Max. motor RPM
    Pk
    Availability
    Price
  • Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm; 17 cfm@85 psi (without trigger)
    Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm; 17 cfm@85 psi (without trigger)
    MFLX07070-60
    07070-60
    Air
    9000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 260,00 €
    1 260,00 €
    1 * 1 ST
       1 260,00 €
    Supplier Article Number: 07070-60
    Local Article Number: MFLB07070-60
    Specifications:
    3A Sanitary approval
    More
    Specifications
    Environmentally Preferable
    Air consumption
    28,9 m³/h (17 ft.³/min)
    Max. viscosity
    1200 cP
    Motor hp
    1/2
    Warranty
    6 Months
    3A Sanitary approval
  • Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm, on/off; 17 cfm@85 psi (with trigger)
    Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm, on/off; 17 cfm@85 psi (with trigger)
    MFLX07070-70
    07070-70
    Air
    9000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 470,00 €
    1 470,00 €
    1 * 1 ST
       1 470,00 €
    Supplier Article Number: 07070-70
    Local Article Number: MFLB07070-70
    Specifications:
    CE Compliant
    3A Sanitary approval
    More
    Specifications
    Environmentally Preferable
    Air consumption
    Max. viscosity
    1200 cP
    Motor hp
    1/2
    Warranty
    6 Months
    CE Compliant
    3A Sanitary approval