Netherlands
Order Entry
Masterflex® Chemically Inert Metering Valves, Avantor®
Masterflex® Chemically Inert Metering Valves, Avantor®
Certificates
undefined
Masterflex® Chemically Inert Metering Valves, Avantor®
Specifications
  • Model:
  • 06394-16
  • 06394-00
  • 06394-02
  • 06394-04
  • Max. pressure:
  • 5,2 bar (75 psi)
  • 5,2 bar (75 psi)
  • 5,2 bar (75 psi)
  • 5,2 bar (75 psi)
  • Max. temperature:
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • Orifice:
  • 6 mm (0,25")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • Wetted materials:
  • PTFE
  • PCTFE and PTFE
  • PCTFE and PTFE
  • PCTFE and PTFE
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Information
  • Catalog #
    Supplier #
    Housing material
    Connection Type
    Inlet port
    Outlet port
    Port Size
    Availability
    Price
  • Chemically Inert Metering Valve, 36 ml/min
    Chemically Inert Metering Valve, 36 ml/min
    MFLX06394-00
    06394-00
    Aluminium
    NPT(F)
    1/8" NPT(F)
    1/8" NPT(F)
    0,125 NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 150,00 €
    150,00 €
    1 * 1 ST
       150,00 €
    Supplier Article Number: 06394-00
    Local Article Number: MFLB06394-00
    Specifications:
    More
    Specifications
    Model
    06394-00
    Max. pressure
    5,2 bar (75 psi)
    Environmentally Preferable
    Max. temperature
    65 °C (150 °F)
    Orifice
    3,18 mm (0,125")
    Wetted materials
    PCTFE and PTFE
    Warranty
    1 year
  • Chemically Inert Metering Valve, 180 ml/min
    Chemically Inert Metering Valve, 180 ml/min
    MFLX06394-02
    06394-02
    Aluminium
    NPT(F)
    1/8" NPT(F)
    1/8" NPT(F)
    0,125 NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 150,00 €
    150,00 €
    1 * 1 ST
       150,00 €
    Supplier Article Number: 06394-02
    Local Article Number: MFLB06394-02
    Specifications:
    More
    Specifications
    Model
    06394-02
    Max. pressure
    5,2 bar (75 psi)
    Environmentally Preferable
    Max. temperature
    65 °C (150 °F)
    Orifice
    3,18 mm (0,125")
    Wetted materials
    PCTFE and PTFE
    Warranty
    1 year
  • Chemically Inert Metering Valve, 1800 ml/min
    Chemically Inert Metering Valve, 1800 ml/min
    MFLX06394-04
    06394-04
    Aluminium
    NPT(F)
    1/8" NPT(F)
    1/8" NPT(F)
    0,125 NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 150,00 €
    150,00 €
    1 * 1 ST
       150,00 €
    Supplier Article Number: 06394-04
    Local Article Number: MFLB06394-04
    Specifications:
    More
    Specifications
    Model
    06394-04
    Max. pressure
    5,2 bar (75 psi)
    Environmentally Preferable
    Max. temperature
    65 °C (150 °F)
    Orifice
    3,18 mm (0,125")
    Wetted materials
    PCTFE and PTFE
    Warranty
    1 year
  • Chemically Inert Metering Valve, 300 L/min Air and 9 L/min Water
    Chemically Inert Metering Valve, 300 L/min Air and 9 L/min Water
    MFLX06394-16
    06394-16
    PTFE
    NPT(F)
    3/8" NPT(F)
    3/8" NPT(F)
    0,375 NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 191,00 €
    191,00 €
    1 * 1 ST
       191,00 €
    Supplier Article Number: 06394-16
    Local Article Number: MFLB06394-16
    Specifications:
    More
    Specifications
    Model
    06394-16
    Max. pressure
    5,2 bar (75 psi)
    Environmentally Preferable
    Max. temperature
    65 °C (150 °F)
    Orifice
    6 mm (0,25")
    Wetted materials
    PTFE
    Warranty
    1 year