FDA-Compliant Viton® Transfer Pump Tubing
Supplier: Avantor Fluid Handling
Certificates
About this item
FDA compliant for food contact.
- Global market leader in range of formulations for most extensive chemical compatibility
Applications: FDA compliant food or laboratory applications, acid and solvent transfer, high temperature work.
Characteristics: Good chemical resistance, opaque white.
Certification: FDA compliant.
Temperature range: -25 to 400 °F(-32 to 204 °C)
Sterilize: Circulating hot air oven at 480 °F (249 °C)
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX06435-05
- MFLX06435-06
- MFLX06435-03
- MFLX06435-04
- MFLX06435-07
- MFLX06435-08
- MFLX06435-09
- MFLX06435-01
- MFLX06435-02
- Clarity:
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Max. pressure:
- 0,9997402 bar (14,5 psi)
- 0,9997402 bar (14,5 psi)
- 0,9997402 bar (14,5 psi)
- 0,9997402 bar (14,5 psi)
- 0,9997402 bar (14,5 psi)
- 0,9997402 bar (14,5 psi)
- 0,9997402 bar (14,5 psi)
- 0,9997402 bar (14,5 psi)
- 0,9997402 bar (14,5 psi)
- Max. temperature:
- 205 °C (400 °F)
- 205 °C (400 °F)
- 205 °C (400 °F)
- 205 °C (400 °F)
- 205 °C (400 °F)
- 205 °C (400 °F)
- 205 °C (400 °F)
- 205 °C (400 °F)
- 205 °C (400 °F)
- Min. temperature:
- –32 °C (–25 °F)
- –32 °C (–25 °F)
- –32 °C (–25 °F)
- –32 °C (–25 °F)
- –32 °C (–25 °F)
- –32 °C (–25 °F)
- –32 °C (–25 °F)
- –32 °C (–25 °F)
- –32 °C (–25 °F)
- Regulatory approvals:
- FDA
- FDA
- FDA
- FDA
- FDA
- FDA
- FDA
- FDA
- FDA
- Sterilisation method autoclave:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Sterilisation method EtO:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Working pressure:
- 14,5 psi
- 14,5 psi
- 14,5 psi
- 14,5 psi
- 14,5 psi
- 14,5 psi
- 14,5 psi
- 14,5 psi
- 14,5 psi
Specifications
Product Information
- Catalog #Tubing int.ØWall thicknessTubing ext.ØTubing materialLengthColourPkAvailabilityPrice
- Transfer Pump Tubing, FDA-Compliant Viton®, White, 5/16" ID x 7/16" OD; 25 ftTransfer Pump Tubing, FDA-Compliant Viton®, White, 5/16" ID x 7/16" OD; 25 ft7,9375 mm (0,3125")1,6 mm11,1 mm (0,4375")Viton® FDA-compliant7,6 mWhite25 FeetLog in for availability1 * 25 Voet311,00 €Local Article Number: MFLA06435-05Specifications:CE certificate
More
Specifications
Environmentally PreferableClarityOpaqueMax. pressure0,9997402 bar (14,5 psi)Max. temperature205 °C (400 °F)Min. temperature–32 °C (–25 °F)Regulatory approvalsFDASterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNoWorking pressure14,5 psiCE certificate - Transfer Pump Tubing, FDA-Compliant Viton®, White, 3/8" ID x 1/2" OD; 25 ftTransfer Pump Tubing, FDA-Compliant Viton®, White, 3/8" ID x 1/2" OD; 25 ft9,525 mm (0,375")1,6 mm12,7 mm (0,5")Viton® FDA-compliant7,6 mWhite25 FeetLog in for availability1 * 25 Voet363,00 €Local Article Number: MFLA06435-06Specifications:CE certificate
More
Specifications
Environmentally PreferableClarityOpaqueMax. pressure0,9997402 bar (14,5 psi)Max. temperature205 °C (400 °F)Min. temperature–32 °C (–25 °F)Regulatory approvalsFDASterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNoWorking pressure14,5 psiCE certificate - Transfer Pump Tubing, FDA-Compliant Viton®, White, 3/16" ID x 1/4" OD; 25 ftTransfer Pump Tubing, FDA-Compliant Viton®, White, 3/16" ID x 1/4" OD; 25 ft4,7625 mm (0,1875")0,8 mm6,4 mm (0,25")Viton® FDA-compliant7,6 mWhite25 FeetLog in for availability1 * 25 Voet125,00 €Local Article Number: MFLA06435-03Specifications:CE certificate
More
Specifications
Environmentally PreferableClarityOpaqueMax. pressure0,9997402 bar (14,5 psi)Max. temperature205 °C (400 °F)Min. temperature–32 °C (–25 °F)Regulatory approvalsFDASterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNoWorking pressure14,5 psiCE certificate - Transfer Pump Tubing, FDA-Compliant Viton®, White, 1/4" ID x 5/16" OD; 25 ftTransfer Pump Tubing, FDA-Compliant Viton®, White, 1/4" ID x 5/16" OD; 25 ft6,35 mm (0,25")0,8 mm7,9 mm (0,3125")Viton® FDA-compliant7,6 mWhite25 FeetLog in for availability1 * 25 Voet142,00 €Local Article Number: MFLA06435-04Specifications:CE certificate
More
Specifications
Environmentally PreferableClarityOpaqueMax. pressure0,9997402 bar (14,5 psi)Max. temperature205 °C (400 °F)Min. temperature–32 °C (–25 °F)Regulatory approvalsFDASterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNoWorking pressure14,5 psiCE certificate - Transfer Pump Tubing, FDA-Compliant Viton®, White, 1/2" ID x 5/8" OD; 25 ftTransfer Pump Tubing, FDA-Compliant Viton®, White, 1/2" ID x 5/8" OD; 25 ft12,7 mm (0,5")1,6 mm15,9 mm (0,625")Viton® FDA-compliant7,6 mWhite25 FeetLog in for availability1 * 25 Voet385,00 €Local Article Number: MFLA06435-07Specifications:CE certificate
More
Specifications
Environmentally PreferableClarityOpaqueMax. pressure0,9997402 bar (14,5 psi)Max. temperature205 °C (400 °F)Min. temperature–32 °C (–25 °F)Regulatory approvalsFDASterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNoWorking pressure14,5 psiCE certificate - Transfer Pump Tubing, FDA-Compliant Viton®, White, 5/8" ID x 7/8" OD; 25 ftTransfer Pump Tubing, FDA-Compliant Viton®, White, 5/8" ID x 7/8" OD; 25 ft15,875 mm (0,625")3,175 mm22,2 mm (0,875")Viton® FDA-compliant7,6 mWhite25 FeetLog in for availability1 * 25 Voet851,00 €Local Article Number: MFLA06435-08Specifications:CE certificate
More
Specifications
Environmentally PreferableClarityOpaqueMax. pressure0,9997402 bar (14,5 psi)Max. temperature205 °C (400 °F)Min. temperature–32 °C (–25 °F)Regulatory approvalsFDASterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNoWorking pressure14,5 psiCE certificate - Transfer Pump Tubing, FDA-Compliant Viton®, White, 3/4" ID x 1" OD; 25 ftTransfer Pump Tubing, FDA-Compliant Viton®, White, 3/4" ID x 1" OD; 25 ft19,05 mm (0,75")3,175 mm25,4 mm (1")Viton® FDA-compliant7,6 mWhite25 FeetLog in for availability1 * 25 Voet962,00 €Local Article Number: MFLA06435-09Specifications:CE certificate
More
Specifications
Environmentally PreferableClarityOpaqueMax. pressure0,9997402 bar (14,5 psi)Max. temperature205 °C (400 °F)Min. temperature–32 °C (–25 °F)Regulatory approvalsFDASterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNoWorking pressure14,5 psiCE certificate - Transfer Pump Tubing, FDA-Compliant Viton®, White, 1/16" ID x 1/8" OD; 25 ftTransfer Pump Tubing, FDA-Compliant Viton®, White, 1/16" ID x 1/8" OD; 25 ft15,875 mm (0,625")0,8 mm3,2 mm (0,125")Viton® FDA-compliant7,6 mWhite25 FeetLog in for availability1 * 25 Voet62,70 €Local Article Number: MFLA06435-01Specifications:CE certificate
More
Specifications
Environmentally PreferableClarityOpaqueMax. pressure0,9997402 bar (14,5 psi)Max. temperature205 °C (400 °F)Min. temperature–32 °C (–25 °F)Regulatory approvalsFDASterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNoWorking pressure14,5 psiCE certificate - Transfer Pump Tubing, FDA-Compliant Viton®, White, 1/8" ID x 3/16" OD; 25 ftTransfer Pump Tubing, FDA-Compliant Viton®, White, 1/8" ID x 3/16" OD; 25 ft3,175 mm (0,125")0,8 mm4,8 mm (0,1875")Viton® FDA-compliant7,6 mWhite25 FeetLog in for availability1 * 25 Voet81,90 €Local Article Number: MFLA06435-02Specifications:CE certificate
More
Specifications
Environmentally PreferableClarityOpaqueMax. pressure0,9997402 bar (14,5 psi)Max. temperature205 °C (400 °F)Min. temperature–32 °C (–25 °F)Regulatory approvalsFDASterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNoWorking pressure14,5 psiCE certificate
Frequently Bought Together