Netherlands
Order Entry
Low IgG FBS, Corning®
Low IgG FBS, Corning®
Catalog # MDTC35-073-CV
Supplier:  Corning
undefined
Low IgG FBS, Corning®
Catalog # MDTC35-073-CV
Supplier:  Corning
Supplier Article Number: 35-073-CV
Local Article Number: MDTC35-073-CV
Price
2 080,00 €
Log in for availability
Unit of Measure 1 * 500 mL
Specifications
  • Description:
    FBS, low IgG
  • Pk:
    500 mL
Specifications
About this item
Low IgG FBS is used in IgG Antibody production, in the propagation of organisms that are normally hindered by the presence of IgGs, or any project in which downstream results would be hindered by exogenous IgGs.
  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Low IgG FBS has reduced levels of IgG which may be required for commercial antibody production or specialised organism propagation
  • Low IgG FBS is sourced from US herds