Netherlands
Order Entry
Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
undefined
Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
  • Inlet connection:
  • None
  • Tube ext.Ø, outlet:
  • 1,4 cm (0,5625")
  • 1,4 cm (0,5625")
  • 1,4 cm (0,5625")
  • Tube length, inlet:
  • 45,7 cm (18")
  • Tube length, outlet:
  • 121,9 cm (48")
  • 121,9 cm (48")
  • 152,4 cm (60")
  • Wetted materials:
  • PVC barrel and hose, LDPE tube
  • PVC barrel and hose, LDPE tube
  • PVC barrel, shaft, hose
  •  
    Flow rate
    Container adapter
    Pk
  • Hand-operated chemical siphon/drum pump, 12 strokes/gallon
    Hand-operated chemical siphon/drum pump, 12 strokes/gallon
    70607-12
    4 strokes/L (12 strokes/gal.)
    2" IPS
    1
    1 * 1 ST
    1 * 1 ST - 125,00 €
    125,00 €
    1 * 1 ST
       125,00 €
     : 70607-12
     : MFLB70607-12
    Specifications
    Environmentally Preferable
    Inlet connection
    Tube ext.Ø, outlet
    1,4 cm (0,5625")
    Tube length, inlet
    Tube length, outlet
    121,9 cm (48")
    Wetted materials
    PVC barrel and hose, LDPE tube
  • Hand-operated chemical siphon/drum pump, 26 strokes/gallon
    Hand-operated chemical siphon/drum pump, 26 strokes/gallon
    70607-10
    7 strokes/L (26 strokes/gal.)
    2" IPS
    1
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
     : 70607-10
     : MFLB70607-10
    Specifications
    Environmentally Preferable
    Inlet connection
    Tube ext.Ø, outlet
    1,4 cm (0,5625")
    Tube length, inlet
    45,7 cm (18")
    Tube length, outlet
    121,9 cm (48")
    Wetted materials
    PVC barrel and hose, LDPE tube
  • Hand-operated water and chemical siphon/drum pump, 32 strokes/gallon
    Hand-operated water and chemical siphon/drum pump, 32 strokes/gallon
    70607-00
    9 strokes/L (32 strokes/gal.)
    None
    1
    1 * 1 ST
    1 * 1 ST - 64,90 €
    64,90 €
    1 * 1 ST
       64,90 €
     : 70607-00
     : MFLB70607-00
    Specifications
    Environmentally Preferable
    Inlet connection
    None
    Tube ext.Ø, outlet
    1,4 cm (0,5625")
    Tube length, inlet
    Tube length, outlet
    152,4 cm (60")
    Wetted materials
    PVC barrel, shaft, hose