Netherlands
Order Entry
Masterflex® Low Pressure-Drop Liquid Flow Switches, Avantor®
Masterflex® Low Pressure-Drop Liquid Flow Switches, Avantor®
Certificates
undefined
Masterflex® Low Pressure-Drop Liquid Flow Switches, Avantor®
Specifications
  • Max. operating temperature:
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • Warranty:
  • 1 year
  • 1 year
Specifications
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Normally closed switch for liquids; 0,5 gal./min
    Normally closed switch for liquids; 0,5 gal./min
    MFLX32776-20
    32776-20
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 230,00 €
    230,00 €
    1 * 1 ST
       230,00 €
    Specifications:
    CE Compliant
    CSA Certified
    UL recognized
    More
    Specifications
    Environmentally Preferable
    Max. operating temperature
    100 °C (212 °F)
    Warranty
    1 year
    CE Compliant
    CSA Certified
    UL recognized
  • Normally closed switch for liquids; 5,0 gal./min
    Normally closed switch for liquids; 5,0 gal./min
    MFLX32776-44
    32776-44
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 230,00 €
    230,00 €
    1 * 1 ST
       230,00 €
    Specifications:
    CSA Certified
    More
    Specifications
    Environmentally Preferable
    Max. operating temperature
    100 °C (212 °F)
    Warranty
    1 year
    CSA Certified