Masterflex® Plastic Ball Valves, Two-way Right Angle, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Nonmetallic wetted parts ideal for water and mild aqueous solutions.
- Maximum pressure: 125 psi (8.6 bar)
- Maximum temperature: 140 °F (60 °C)
Ball valves are designed primarily for on/off service.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX01378-50
- MFLX01378-54
- MFLX01379-22
- MFLX01378-56
- MFLX01378-64
- MFLX01379-23
- MFLX01379-24
- MFLX01378-46
- MFLX01378-48
- MFLX01378-52
- MFLX01378-58
- MFLX01379-15
- MFLX01379-16
- MFLX01378-72
- MFLX01378-74
- MFLX01378-76
- MFLX01378-60
- MFLX01378-66
- MFLX01378-68
- MFLX01378-70
- MFLX01379-27
- MFLX01379-28
- MFLX01379-30
- MFLX01379-17
- MFLX01379-18
- MFLX01379-19
- MFLX01379-20
- Model:
- 01378-50
- 01378-54
- 01379-22
- 01378-56
- 01378-64
- 01379-23
- 01379-24
- 01378-46
- 01378-48
- 01378-52
- 01378-58
- 01379-15
- 01379-16
- 01378-72
- 01378-74
- 01378-76
- 01378-60
- 01378-66
- 01378-68
- 01378-70
- 01379-27
- 01379-28
- 01379-30
- 01379-17
- 01379-18
- 01379-19
- 01379-20
- Interne Ø slangen:
- 0,25"
- 0,25"
- 0,25"
- 0,25"
- 0,375"
- 0,375"
- Tubing ext.Ø:
- 0,25"
- 0,25"
- 0,25"
- 0,375"
- 0,375"
- 0,375"
- 0,375"
- 0,375"
- 0,375"
- 0,375"
- 0,25"
- 0,25"
- 0,25"
- 0,25"
- Max. druk:
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- 125 psi
- Maximumtemperatuur:
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- Orifice:
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- 11,0998 mm (0,437")
- Wetted materials:
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- PVC
- Garantie:
- 6 Months
- 6 Months
- 6 Months
- 6 Months
- 6 Months
- 6 Months
- 6 Months
- 6 Months
- 6 Months
- 6 Months
- 6 Months
- 6 Months
- 6 Months
- 6 Months
- 6 Months
Specifications
Product Information
- Catalog #Supplier #OmschrijvingVerbindingstypeInlet portOutlet portPoortgrootteCvAvailabilityPrice
- 01378-46Ball valve, 2-way right angled, 1/4" NPT(F), PVC with Buna N sealsNPT(F)1/4" NPT(F)1/4" NPT(F)0,25 NPT(F)7Log in for availability1 * 1 ST26,90 €Supplier Article Number: 01378-46Local Article Number: MFLB01378-46Specifications:
More
Specifications
Model01378-46Environmentally PreferableInterne Ø slangenTubing ext.ØMax. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-48Ball valve, 2-way right angled, 1/4" NPT(F); PVC with EPDM sealsNPT(F)1/4" NPT(F)1/4" NPT(F)0,25 NPT(F)7Log in for availability1 * 1 ST26,90 €Supplier Article Number: 01378-48Local Article Number: MFLB01378-48Specifications:
More
Specifications
Model01378-48Environmentally PreferableInterne Ø slangenTubing ext.ØMax. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-50Ball valve, 2-way right angled, 1/4" NPT(F) x 1/4" NPT(M), PVC with Buna N sealsNPT(F) x NPT(M)1/4" NPT(F)1/4" NPT(M)0,25 NPT(F)7Log in for availability1 * 1 ST26,80 €Supplier Article Number: 01378-50Local Article Number: MFLB01378-50Specifications:
More
Specifications
Model01378-50Environmentally PreferableInterne Ø slangenTubing ext.ØMax. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-52Ball valve, 2-way right angled, 1/4" NPT(F) x 1/4" NPT(M), PVC with EPDM sealsNPT(F) x NPT(M)1/4" NPT(F)1/4" NPT(M)0,25 NPT(F)7Log in for availability1 * 1 ST26,90 €Supplier Article Number: 01378-52Local Article Number: MFLB01378-52Specifications:
More
Specifications
Model01378-52Environmentally PreferableInterne Ø slangenTubing ext.ØMax. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-54Ball valve, 2-way right angled, 1/4" NPT(F) x 1/4" barb, PVC with Buna N sealsNPT(F) x Barbed1/4" NPT(F)1/4" Barb0,25 NPT(F)7Log in for availability1 * 1 ST26,90 €Supplier Article Number: 01378-54Local Article Number: MFLB01378-54Specifications:
More
Specifications
Model01378-54Environmentally PreferableInterne Ø slangen0,25"Tubing ext.ØMax. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-56Ball valve, 2-way right angled, 1/4" NPT(F) x 1/4" barb, PVC with EPDM sealsNPT(F) x Barbed1/4" NPT(F)1/4" Barb0,25 NPT(F)7Log in for availability1 * 1 ST26,90 €Supplier Article Number: 01378-56Local Article Number: MFLB01378-56Specifications:
More
Specifications
Model01378-56Environmentally PreferableInterne Ø slangen0,25"Tubing ext.ØMax. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-58Ball valve, 2-way right angled, 1/4" NPT(M) x 1/4" barb, PVC with Buna N sealsNPT(M) x Barbed1/4" NPT(M)1/4" Barb7Log in for availability1 * 1 ST29,60 €Supplier Article Number: 01378-58Local Article Number: MFLB01378-58Specifications:
More
Specifications
Model01378-58Environmentally PreferableInterne Ø slangen0,25"Tubing ext.ØMax. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-60Ball valve, 2-way right angled, 1/4" NPT(M) x 1/4" barb, PVC with EPDM sealsNPT(M) x Barbed1/4" NPT(M)1/4" Barb7Log in for availability1 * 1 ST29,60 €Supplier Article Number: 01378-60Local Article Number: MFLB01378-60Specifications:
More
Specifications
Model01378-60Environmentally PreferableInterne Ø slangen0,25"Tubing ext.ØMax. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-64Ball valve, 2-way right angled, 1/4" NPT(F) x 1/4" OD, PVC with EPDM sealsNPT(F) x Compression1/4" NPT(F)1/4" compression0,25 NPT(F)7Log in for availability1 * 1 ST26,80 €Supplier Article Number: 01378-64Local Article Number: MFLB01378-64Specifications:
More
Specifications
Model01378-64Environmentally PreferableInterne Ø slangenTubing ext.Ø0,25"Max. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-66Ball valve, 2-way right angled, 3/8" NPT(M) x 3/8" OD, PVC with Buna N sealsNPT(M) x Compression3/8" NPT(M)3/8" compression7Log in for availability1 * 1 ST29,60 €Supplier Article Number: 01378-66Local Article Number: MFLB01378-66Specifications:
More
Specifications
Model01378-66Environmentally PreferableInterne Ø slangenTubing ext.Ø0,375"Max. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-68Ball valve, 2-way right angled, 3/8" NPT(M) x 3/8" OD, PVC with EPDM sealsNPT(M) x Compression3/8" NPT(M)3/8" compression7Log in for availability1 * 1 ST29,60 €Supplier Article Number: 01378-68Local Article Number: MFLB01378-68Specifications:
More
Specifications
Model01378-68Environmentally PreferableInterne Ø slangenTubing ext.Ø0,375"Max. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-70Ball valve, 2-way right angled, 3/8"NPT(F) x 3/8" John Guest, PVC w/Buna N sealsNPT(F) x John Guest3/8" NPT(F)3/8" John Guest0,375 NPT(F)7Log in for availability1 * 1 ST32,40 €Supplier Article Number: 01378-70Local Article Number: MFLB01378-70Specifications:
More
Specifications
Model01378-70Environmentally PreferableInterne Ø slangenTubing ext.Ø0,375"Max. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-72Ball valve, 2-way right angled, 3/8"NPT(F) x 3/8" John Guest, PVC w/EPDM sealsNPT(F) x John Guest3/8" NPT(F)3/8" John Guest0,375 NPT(F)7Log in for availability1 * 1 ST31,10 €Supplier Article Number: 01378-72Local Article Number: MFLB01378-72Specifications:
More
Specifications
Model01378-72Environmentally PreferableInterne Ø slangenTubing ext.Ø0,375"Max. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-74Ball valve, 2-way right angled, 3/8"NPT(M) x 3/8" John Guest, PVC w/Buna N sealsNPT(M) x John Guest3/8" NPT(M)3/8" John Guest7Log in for availability1 * 1 ST32,50 €Supplier Article Number: 01378-74Local Article Number: MFLB01378-74Specifications:
More
Specifications
Model01378-74Environmentally PreferableInterne Ø slangenTubing ext.Ø0,375"Max. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01378-76Ball valve, 2-way right angled, 3/8"NPT(M) x 3/8" John Guest, PVC w/EPDM sealsNPT(M) x John Guest3/8" NPT(M)3/8" John Guest7Log in for availability1 * 1 ST32,40 €Supplier Article Number: 01378-76Local Article Number: MFLB01378-76Specifications:
More
Specifications
Model01378-76Environmentally PreferableInterne Ø slangenTubing ext.Ø0,375"Max. druk125 psiMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie6 Months - 01379-15Ball valve, 2-way right angled, BUNA, 1/4" NPT(M) x 1/4" compressionNPT(M) x Compression1/4" NPT(M)1/4" compression7Log in for availability1 * 1 ST26,90 €Supplier Article Number: 01379-15Local Article Number: MFLB01379-15Specifications:
More
Specifications
Model01379-15Environmentally PreferableInterne Ø slangenTubing ext.Ø0,25"Max. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie - 01379-16Ball valve, 2-way right angled, EPDM, 1/4" NPT(M) x 1/4" compressionNPT(M) x Compression1/4" NPT(M)1/4" compression7Log in for availability1 * 1 ST26,80 €Supplier Article Number: 01379-16Local Article Number: MFLB01379-16Specifications:
More
Specifications
Model01379-16Environmentally PreferableInterne Ø slangenTubing ext.Ø0,25"Max. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie - 01379-17Ball valve, 2-way right angled, BUNA, 1/4" NPT(F) x 1/4" John GuestNPT(F) x John Guest1/4" NPT(F)1/4" John Guest0,25 NPT(F)7Log in for availability1 * 1 ST32,50 €Supplier Article Number: 01379-17Local Article Number: MFLB01379-17Specifications:
More
Specifications
Model01379-17Environmentally PreferableInterne Ø slangenTubing ext.Ø0,25"Max. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie - 01379-18Ball valve, 2-way right angled, EPDM, 1/4" NPT(F) x 1/4" John GuestNPT(F) x John Guest1/4" NPT(F)1/4" John Guest0,25 NPT(F)7Log in for availability1 * 1 ST29,10 €Supplier Article Number: 01379-18Local Article Number: MFLB01379-18Specifications:
More
Specifications
Model01379-18Environmentally PreferableInterne Ø slangenTubing ext.Ø0,25"Max. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie - 01379-19Ball valve, 2-way right angled, 1/4"NPT(M) x 1/4" John Guest, PVC w/Buna N sealsNPT(M) x John Guest1/4" NPT(M)1/4" John Guest7Log in for availability1 * 1 ST32,40 €Supplier Article Number: 01379-19Local Article Number: MFLB01379-19Specifications:
More
Specifications
Model01379-19Environmentally PreferableInterne Ø slangenTubing ext.Ø0,25"Max. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie - 01379-20Ball valve, 2-way right angled, 1/4" NPT(M) x 1/4" John Guest, PVC w/EPDM sealsNPT(M) x John Guest1/4" NPT(M)1/4" John Guest7Log in for availability1 * 1 ST32,40 €Supplier Article Number: 01379-20Local Article Number: MFLB01379-20Specifications:
More
Specifications
Model01379-20Environmentally PreferableInterne Ø slangenTubing ext.Ø0,25"Max. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie - 01379-22Ball valve, 2-way right angled, EPDM, 3/8" NPT(F)NPT(F)3/8" NPT(F)3/8" NPT(F)0,375 NPT(F)7Log in for availability1 * 1 ST26,90 €Supplier Article Number: 01379-22Local Article Number: MFLB01379-22Specifications:
More
Specifications
Model01379-22Environmentally PreferableInterne Ø slangenTubing ext.ØMax. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie - 01379-23Ball valve, 2-way right angled, BUNA, 3/8" NPT(F) x 3/8" NPT(M)NPT(F) x NPT(M)3/8" NPT(F)3/8" NPT(M)0,375 NPT(F)7Log in for availability1 * 1 ST26,90 €Supplier Article Number: 01379-23Local Article Number: MFLB01379-23Specifications:
More
Specifications
Model01379-23Environmentally PreferableInterne Ø slangenTubing ext.ØMax. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie - 01379-24Ball valve, 2-way right angled, EPDM, 3/8" NPT(F) x 3/8" NPT(M)NPT(F) x NPT(M)3/8" NPT(F)3/8" NPT(M)0,375 NPT(F)7Log in for availability1 * 1 ST26,90 €Supplier Article Number: 01379-24Local Article Number: MFLB01379-24Specifications:
More
Specifications
Model01379-24Environmentally PreferableInterne Ø slangenTubing ext.ØMax. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie - 01379-27Ball valve, 2-way right angled, BUNA, 3/8" NPT(M) x 3/8" barbNPT(F) x Barbed3/8" NPT(M)3/8" Barb0,375 NPT(F)7Log in for availability1 * 1 ST26,90 €Supplier Article Number: 01379-27Local Article Number: MFLB01379-27Specifications:
More
Specifications
Model01379-27Environmentally PreferableInterne Ø slangen0,375"Tubing ext.ØMax. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie - 01379-28Ball valve, 2-way right angled, EPDM, 3/8" NPT(M) x 3/8" barbNPT(F) x Barbed3/8" NPT(M)3/8" Barb0,375 NPT(F)7Log in for availability1 * 1 ST26,90 €Supplier Article Number: 01379-28Local Article Number: MFLB01379-28Specifications:
More
Specifications
Model01379-28Environmentally PreferableInterne Ø slangen0,375"Tubing ext.ØMax. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie - 01379-30Ball valve, 2-way right angled, EPDM, 3/8" NPT(F) x 3/8" compressionNPT(F) x Compression3/8" NPT(F)3/8" compression0,375 NPT(F)7Log in for availability1 * 1 ST26,80 €Supplier Article Number: 01379-30Local Article Number: MFLB01379-30Specifications:
More
Specifications
Model01379-30Environmentally PreferableInterne Ø slangenTubing ext.Ø0,375"Max. drukMaximumtemperatuur60 °C (140 °F)Orifice11,0998 mm (0,437")Wetted materialsPVCGarantie