About this item
Ensure a sterile connection.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- One disposable connector for steam sterilization and microbial process
- Reduces down time and cleaning expenses
As part of a single-use system, these CPC® sanitary Steam-in-Place disposable connectors enhance the sterility of your bioprocessing line. The innovative product design allows for a one-step process for sterilization and media transfer while eliminating dead spots, reducing the number of connections, and eliminating the need for a laminar flow hood. Terminations on these connectors are set to industry standards for bio-reactors and popular sizes of flexible tubing. All wetted surfaces meet requirements for USP Class VI, and physicochemical tests. All units are made of animal-free polysulfone resin.
The single-cycle steam-thru connectors feature a three-port design that allows steam to pass directly through the power ports to 'steam on' to stainless equipment. Once the SIP cycle is completed, remove the tear-away sleeve to actuate the valve and create a sterile flow path. Once actuated, the connection cannot be reversed because valve actuation punctures the internal membrane to allow flow of media. When the process is complete, the connector may be safely disposed.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX31604-18BLK
- MFLX31604-18
- MFLX31309-61BLK
- MFLX31604-20
- MFLX31604-20BLK
- MFLX31604-10BLK
- MFLX31604-10
- Max. operating pressure:
- 20 psi
- 20 psi
- 20 psi
- 20 psi
- 20 psi
- 20 psi
- 20 psi
- Max. temperature:
- 40 °C (104 °F)
- 40 °C (104 °F)
- 40 °C (104 °F)
- 40 °C (104 °F)
- 40 °C (104 °F)
- 40 °C (104 °F)
- 40 °C (104 °F)
- Min. temperature:
- 4 °C (39 °F)
- 4 °C (39 °F)
- 4 °C (39 °F)
- 4 °C (39 °F)
- 4 °C (39 °F)
- 4 °C (39 °F)
- 4 °C (39 °F)
- Seal material:
- Platinum-cured Silicone
- Platinum-cured Silicone
- Platinum-cured Silicone
- Platinum-cured Silicone
- Platinum-cured Silicone
- Platinum-cured Silicone
- Platinum-cured Silicone
- Sterilisation:
- Autoclave, gamma, or steam-in-place (SIP)
- Autoclave, gamma, or steam-in-place (SIP)
- Autoclave, gamma, or steam-in-place (SIP)
- Autoclave, gamma, or steam-in-place (SIP)
- Autoclave, gamma, or steam-in-place (SIP)
- Gamma or steam-in-place (SIP)
- Gamma or steam-in-place (SIP)
- Warranty:
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
Specifications
Down pack product
Product Information
- Catalog #Supplier #Barb size/tubing int.Ø (port 3)Connection TypeFitting materialFlow patternSanitary clamp size (Port 1)AvailabilityPrice
- Sanitary Steam-in-Place Connector, Single Cycle, Polycarbonate Sleeve, ¹/₂" ID׳/₄"׳/₄" Sanitary, STC1700500; 1/PKSanitary Steam-in-Place Connector, Single Cycle, Polycarbonate Sleeve, ¹/₂" ID׳/₄"׳/₄" Sanitary, STC1700500; 1/PK31604-18½"Sanitary to BarbPolysulfoneStraight¾"Log in for availability1 * 1 ST308,00 €Supplier Article Number: 31604-18Local Article Number: MFLB31604-18Specifications:
More
Specifications
Environmentally PreferableMax. operating pressure20 psiMax. temperature40 °C (104 °F)Min. temperature4 °C (39 °F)Seal materialPlatinum-cured SiliconeSterilisationAutoclave, gamma, or steam-in-place (SIP)Warranty3 Months - Sanitary Steam-in-Place Connector, Single Cycle, Polycarbonate Sleeve, ³/₈" ID׳/₄"׳/₄" Sanitary, STC1700600; 1/PKSanitary Steam-in-Place Connector, Single Cycle, Polycarbonate Sleeve, ³/₈" ID׳/₄"׳/₄" Sanitary, STC1700600; 1/PK31604-20⅜"Sanitary to BarbPolysulfoneStraight¾"Log in for availability1 * 1 ST308,00 €Supplier Article Number: 31604-20Local Article Number: MFLB31604-20Specifications:
More
Specifications
Environmentally PreferableMax. operating pressure20 psiMax. temperature40 °C (104 °F)Min. temperature4 °C (39 °F)Seal materialPlatinum-cured SiliconeSterilisationAutoclave, gamma, or steam-in-place (SIP)Warranty3 Months - Sanitary Steam-in-Place Connector, Single Cycle, Polyethylene Sleeve, ¹/₂" ID׳/₄"׳/₄" Sanitary, STC1700000; 1/PKSanitary Steam-in-Place Connector, Single Cycle, Polyethylene Sleeve, ¹/₂" ID׳/₄"׳/₄" Sanitary, STC1700000; 1/PK31604-10½"Sanitary to BarbPolysulfoneStraight¾"Log in for availability1 * 1 ST308,00 €Supplier Article Number: 31604-10Local Article Number: MFLB31604-10Specifications:
More
Specifications
Environmentally PreferableMax. operating pressure20 psiMax. temperature40 °C (104 °F)Min. temperature4 °C (39 °F)Seal materialPlatinum-cured SiliconeSterilisationGamma or steam-in-place (SIP)Warranty3 Months
Bulk product
Product Information
- Catalog #Supplier #Barb size/tubing int.Ø (port 3)Connection TypeFitting materialFlow patternSanitary clamp size (Port 1)AvailabilityPrice
- Sanitary Steam-in-Place Connector, Single Cycle, Polycarbonate Sleeve, ¹/₂" ID׳/₄"׳/₄" Sanitary, STC1700500; 5/PKSanitary Steam-in-Place Connector, Single Cycle, Polycarbonate Sleeve, ¹/₂" ID׳/₄"׳/₄" Sanitary, STC1700500; 5/PK31604-18BLK½"Sanitary to BarbPolysulfoneStraight¾"Log in for availability1 * 5 ST845,00 €Supplier Article Number: 31604-18BLKLocal Article Number: MFLB31604-18BLKSpecifications:
More
Specifications
Environmentally PreferableMax. operating pressure20 psiMax. temperature40 °C (104 °F)Min. temperature4 °C (39 °F)Seal materialPlatinum-cured SiliconeSterilisationAutoclave, gamma, or steam-in-place (SIP)Warranty3 Months - Sanitary Steam-in-Place Connector, Single Cycle, Polycarbonate Sleeve, ³/₈" ID׳/₄"×1-¹/₂" Sanitary, STC1700800; 5/PKSanitary Steam-in-Place Connector, Single Cycle, Polycarbonate Sleeve, ³/₈" ID׳/₄"×1-¹/₂" Sanitary, STC1700800; 5/PK31309-61BLK⅜"Sanitary to BarbPolysulfoneStraight1½"Log in for availability1 * 5 ST789,00 €Supplier Article Number: 31309-61BLKLocal Article Number: MFLB31309-61BLKSpecifications:
More
Specifications
Environmentally PreferableMax. operating pressure20 psiMax. temperature40 °C (104 °F)Min. temperature4 °C (39 °F)Seal materialPlatinum-cured SiliconeSterilisationAutoclave, gamma, or steam-in-place (SIP)Warranty3 Months - Sanitary Steam-in-Place Connector, Single Cycle, Polycarbonate Sleeve, ³/₈" ID׳/₄"׳/₄" Sanitary, STC1700600; 5/PKSanitary Steam-in-Place Connector, Single Cycle, Polycarbonate Sleeve, ³/₈" ID׳/₄"׳/₄" Sanitary, STC1700600; 5/PK31604-20BLK⅜"Sanitary to BarbPolysulfoneStraight¾"Log in for availability1 * 5 ST845,00 €Supplier Article Number: 31604-20BLKLocal Article Number: MFLB31604-20BLKSpecifications:
More
Specifications
Environmentally PreferableMax. operating pressure20 psiMax. temperature40 °C (104 °F)Min. temperature4 °C (39 °F)Seal materialPlatinum-cured SiliconeSterilisationAutoclave, gamma, or steam-in-place (SIP)Warranty3 Months - Sanitary Steam-in-Place Connector, Single Cycle, Polyethylene Sleeve, ¹/₂" ID׳/₄"׳/₄" Sanitary, STC1700000; 5/PKSanitary Steam-in-Place Connector, Single Cycle, Polyethylene Sleeve, ¹/₂" ID׳/₄"׳/₄" Sanitary, STC1700000; 5/PK31604-10BLK½"Sanitary to BarbPolysulfoneStraight¾"Log in for availability1 * 5 ST845,00 €Supplier Article Number: 31604-10BLKLocal Article Number: MFLB31604-10BLKSpecifications:
More
Specifications
Environmentally PreferableMax. operating pressure20 psiMax. temperature40 °C (104 °F)Min. temperature4 °C (39 °F)Seal materialPlatinum-cured SiliconeSterilisationGamma or steam-in-place (SIP)Warranty3 Months