Netherlands
Order Entry
Wire Mesh Overscreens for Sanitary Strainers
Wire Mesh Overscreens for Sanitary Strainers
Certificates
undefined
Wire Mesh Overscreens for Sanitary Strainers
Specifications
  • Filter type:
  • Strainer
  • Strainer
  • Strainer
  • Strainer
  • Strainer
  • Strainer
  • Length:
  • 27,62 cm (10,875")
  • 27,62 cm (10,875")
  • 27,62 cm (10,875")
  • 27,62 cm (10,875")
  • 27,62 cm (10,875")
  • 27,62 cm (10,875")
  • Ø:
  • 7,62 cm (3")
  • 7,62 cm (3")
  • 7,62 cm (3")
  • 7,62 cm (3")
  • 7,62 cm (3")
  • 7,62 cm (3")
Specifications
Product Information
  • Catalog #
    Supplier #
    Mesh size
    Material
    Micron nominal
    Pk
    Availability
    Price
  • Mesh Overscreen for Sanitary Strainer 3" dia; 20 x 20 Mesh
    Mesh Overscreen for Sanitary Strainer 3" dia; 20 x 20 Mesh
    MFLX29590-31
    29590-31
    20
    Stainless steel 316
    864 µm
    1
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 182,00 €
    182,00 €
    1 * 1 ST
       182,00 €
    Supplier Article Number: 29590-31
    Local Article Number: MFLB29590-31
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Filter type
    Strainer
    Length
    27,62 cm (10,875")
    Ø
    7,62 cm (3")
  • Mesh Overscreen for Sanitary Strainer 3" dia; 40 x 40 Mesh
    Mesh Overscreen for Sanitary Strainer 3" dia; 40 x 40 Mesh
    MFLX29590-33
    29590-33
    40
    Stainless steel 316
    381 µm
    1
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 183,00 €
    183,00 €
    1 * 1 ST
       183,00 €
    Supplier Article Number: 29590-33
    Local Article Number: MFLB29590-33
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Filter type
    Strainer
    Length
    27,62 cm (10,875")
    Ø
    7,62 cm (3")
  • Mesh Overscreen for Sanitary Strainer 3" dia; 60 x 60 Mesh
    Mesh Overscreen for Sanitary Strainer 3" dia; 60 x 60 Mesh
    MFLX29590-35
    29590-35
    40
    Stainless steel 316
    234 µm
    1
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 183,00 €
    183,00 €
    1 * 1 ST
       183,00 €
    Supplier Article Number: 29590-35
    Local Article Number: MFLB29590-35
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Filter type
    Strainer
    Length
    27,62 cm (10,875")
    Ø
    7,62 cm (3")
  • Mesh Overscreen for Sanitary Strainer 3" dia; 80 x 80 Mesh
    Mesh Overscreen for Sanitary Strainer 3" dia; 80 x 80 Mesh
    MFLX29590-36
    29590-36
    60
    Stainless steel 316
    178 µm
    1
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 183,00 €
    183,00 €
    1 * 1 ST
       183,00 €
    Supplier Article Number: 29590-36
    Local Article Number: MFLB29590-36
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Filter type
    Strainer
    Length
    27,62 cm (10,875")
    Ø
    7,62 cm (3")
  • Mesh Overscreen for Sanitary Strainer 3" dia; 100 x 100 Mesh
    Mesh Overscreen for Sanitary Strainer 3" dia; 100 x 100 Mesh
    MFLX29590-37
    29590-37
    100
    Stainless steel 316
    140 µm
    1
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 238,00 €
    238,00 €
    1 * 1 ST
       238,00 €
    Supplier Article Number: 29590-37
    Local Article Number: MFLB29590-37
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Filter type
    Strainer
    Length
    27,62 cm (10,875")
    Ø
    7,62 cm (3")
  • Mesh Overscreen for Sanitary Strainer 3" dia; 150 x 150 Mesh
    Mesh Overscreen for Sanitary Strainer 3" dia; 150 x 150 Mesh
    MFLX29590-39
    29590-39
    100
    Stainless steel 316
    104 µm
    1
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 239,00 €
    239,00 €
    1 * 1 ST
       239,00 €
    Supplier Article Number: 29590-39
    Local Article Number: MFLB29590-39
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Filter type
    Strainer
    Length
    27,62 cm (10,875")
    Ø
    7,62 cm (3")