Netherlands
Order Entry
PrimeLock® High-Purity PFA Fittingss Accessories
PrimeLock® High-Purity PFA Fittingss Accessories
PrimeLock® High-Purity PFA Fittingss Accessories
  • Compression size / tube ext.Ø (port 1):
  • 1"
  • 1"
  • 1"
  • Connection type:
  • Flare Installation Insert
  • Flare
  • Flare Wrench
  • Max. operating pressure:
  • 120 psi at 70 °F (21 °C)
  • 120 psi at 70 °F (21 °C)
  • Max. temperature:
  • 200 °C (392 °F)
  • 200 °C (392 °F)
  • Min. temperature:
  • –18 °C (0 °F)
  • –18 °C (0 °F)
  • Warranty:
  • 2 months
  • 2 months
  • 2 months
  •  
    Colour
    Fitting material
    Pk
  • Installation Insert, PFA, 1" OD tubing; 10/pk
    Installation Insert, PFA, 1" OD tubing; 10/pk
    30491-95
    Natural
    PFA
    10
    1 * 10 ST
    1 * 10 ST - 187,00 €
    187,00 €
    1 * 10 ST
       187,00 €
     : 30491-95
     : MFLB30491-95
    2 month warranty
    Accessories
    Specifications
    Environmentally Preferable
    Compression size / tube ext.Ø (port 1)
    1"
    Connection type
    Flare Installation Insert
    Max. operating pressure
    120 psi at 70 °F (21 °C)
    Max. temperature
    200 °C (392 °F)
    Min. temperature
    –18 °C (0 °F)
    Warranty
    2 months
    2 month warranty
  • High-Purity Fitting, nut, standard, 1" OD tubing; 1/ea
    High-Purity Fitting, nut, standard, 1" OD tubing; 1/ea
    30492-02
    Natural
    PFA
    1
    1 * 1 ST
    1 * 1 ST - 37,20 €
    37,20 €
    1 * 1 ST
       37,20 €
     : 30492-02
     : MFLB30492-02
    2 month warranty
    Accessories
    Specifications
    Environmentally Preferable
    Compression size / tube ext.Ø (port 1)
    1"
    Connection type
    Flare
    Max. operating pressure
    120 psi at 70 °F (21 °C)
    Max. temperature
    200 °C (392 °F)
    Min. temperature
    –18 °C (0 °F)
    Warranty
    2 months
    2 month warranty
  • Installation Wrench, 1" OD tubing; 1/ea
    Installation Wrench, 1" OD tubing; 1/ea
    30493-43
    Natural
    PFA
    1
    1 * 1 ST
    1 * 1 ST - 26,00 €
    26,00 €
    1 * 1 ST
       26,00 €
     : 30493-43
     : MFLB30493-43
    2 month warranty
    Accessories
    Specifications
    Environmentally Preferable
    Compression size / tube ext.Ø (port 1)
    1"
    Connection type
    Flare Wrench
    Max. operating pressure
    Max. temperature
    Min. temperature
    Warranty
    2 months
    2 month warranty