Netherlands
Order Entry
Masterflex® Barbed High Flow+ Male Quick-Disconnect Fittings, Avantor®
Masterflex® Barbed High Flow+ Male Quick-Disconnect Fittings, Avantor®
Certificates
undefined
Masterflex® Barbed High Flow+ Male Quick-Disconnect Fittings, Avantor®
Specifications
  • O-Ring material:
  • Silicone
  • Silicone
  • Sterilisation:
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Valve spring material:
  • Stainless Steel
  • Stainless Steel
  • Regulatory approvals:
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
Specifications
Product Information
  • Catalog #
    Type
    Barb size/tubing int.Ø (port 1)
    Connection Type
    Fitting material
    Flow pattern
    Pk
    Availability
    Price
  • High Flow+, 1" Barbed Male, Valved Flow, PVDF Housing, Silicone O-Ring, Stainless Spring
    High Flow+, 1" Barbed Male, Valved Flow, PVDF Housing, Silicone O-Ring, Stainless Spring
    MFLX40250-98
    Valved
    1"
    Quick-Disconnect to Barb
    PVDF
    Straight
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 982,00 €
    982,00 €
    1 * 10 ST
       982,00 €
    Specifications:
    More
    Specifications
    Environmentally Preferable
    O-Ring material
    Silicone
    Sterilisation
    Gamma irradiation, EtO, autoclave
    Valve spring material
    Stainless Steel
    Regulatory approvals
    FDA;USP
  • High Flow+, 1" Barbed Male, Valved Flow, Polypropylene Housing, Silicone O-Ring, Stainless Spring
    High Flow+, 1" Barbed Male, Valved Flow, Polypropylene Housing, Silicone O-Ring, Stainless Spring
    MFLX40250-83
    Valved
    1"
    Quick-Disconnect to Barb
    Polypropylene
    Straight
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 679,00 €
    679,00 €
    1 * 10 ST
       679,00 €
    Specifications:
    More
    Specifications
    Environmentally Preferable
    O-Ring material
    Silicone
    Sterilisation
    Gamma irradiation, EtO, autoclave
    Valve spring material
    Stainless Steel
    Regulatory approvals
    FDA;USP
  • High Flow+, 1" Barbed Male, Open Flow, Polypropylene Housing
    High Flow+, 1" Barbed Male, Open Flow, Polypropylene Housing
    MFLX40250-67
    Open
    1"
    Quick-Disconnect to Barb
    Polypropylene
    Straight
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 400,00 €
    400,00 €
    1 * 10 ST
       400,00 €
    Specifications:
    More
    Specifications
    Environmentally Preferable
    O-Ring material
    Sterilisation
    Gamma irradiation, EtO, autoclave
    Valve spring material
    Regulatory approvals
    FDA;USP
  • High Flow+, 1" Barbed Male, Open Flow, PVDF Housing
    High Flow+, 1" Barbed Male, Open Flow, PVDF Housing
    MFLX40250-75
    Open
    1"
    Quick-Disconnect to Barb
    PVDF
    Straight
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 594,00 €
    594,00 €
    1 * 10 ST
       594,00 €
    Specifications:
    More
    Specifications
    Environmentally Preferable
    O-Ring material
    Sterilisation
    Gamma irradiation, EtO, autoclave
    Valve spring material
    Regulatory approvals
    FDA;USP