Netherlands
Order Entry
Paliperidone Related Compounds, Reference Standard
Paliperidone Related Compounds, Reference Standard
Leverancier:  USP
Certificaten
Paliperidone Related Compounds, Reference Standard
Leverancier:  USP
Specificaties
  • CAS No.:
    84163-13-3
  • Synonym(s):
    6-fluoro-3-(piperidin-4-yl)benzoisoxazole hydrochloride
  • Formula:
    C₁₂H₁₄ClFN₂O
  • Description:
    Paliperidone Related Compound B
Specificaties
Productinformatie
  • Catalog #
    Supplier #
    Standards
    Pack type
    Pk
    Beschikbaarheid
    Prijs
  • Paliperidone Related Compound A
    Paliperidone Related Compound A
    1491818.
    1491818
    USP reference standard
    Vial
    20 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 20 mg
    1 * 20 mg - 1 080,00 €
    1 080,00 €
    1 * 20 mg
       1 080,00 €
    Supplier Article Number: 1491818
    Local Article Number: USPH1491818
  • Paliperidone Related Compound B
    Paliperidone Related Compound B
    1491820.
    1491820
    USP reference standard
    Vial
    20 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 20 mg
    1 * 20 mg - 1 140,00 €
    1 140,00 €
    1 * 20 mg
       1 140,00 €
    Supplier Article Number: 1491820
    Local Article Number: USPH1491820
  • Paliperidone Related Compound C
    Paliperidone Related Compound C
    1491831.
    1491831
    USP reference standard
    Vial
    20 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 20 mg
    1 * 20 mg - 1 200,00 €
    1 200,00 €
    1 * 20 mg
       1 200,00 €
    Supplier Article Number: 1491831
    Local Article Number: USPH1491831
  • Paliperidone Related Compound D
    Paliperidone Related Compound D
    1491842.
    1491842
    USP reference standard
    Vial
    20 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 20 mg
    1 * 20 mg - 1 140,00 €
    1 140,00 €
    1 * 20 mg
       1 140,00 €
    Supplier Article Number: 1491842
    Local Article Number: USPH1491842