Netherlands
Order Entry
CultiMaxx Shelving Systems for Heracell VIOS 250i and Heracell Vios 250i CR CTS Series CO₂ Incubators
CultiMaxx Shelving Systems for Heracell VIOS 250i and Heracell Vios 250i CR CTS Series CO₂ Incubators
undefined
CultiMaxx Shelving Systems for Heracell VIOS 250i and Heracell Vios 250i CR CTS Series CO₂ Incubators
  • Catalog No:
  • 390-1671
  • 390-1693
  • 390-1694
  • Capacity:
  • 85 L
  • Temperature range:
  • –50...–86 °C
Shelving System for Nunc Cell Factory System with 4 layers
  •  
    For
    W×D×H int.
    W×D×H ext.
    Pk
  • Set of 5 shelves to support up to ten 4- or 5-layer culture vessels, such as Thermo Scientific™ Nunc™ Cell Factory™ Systems
    Set of 5 shelves to support up to ten 4- or 5-layer culture vessels, such as Thermo Scientific™ Nunc™ Cell Factory™ Systems
    390-1693
    50166268
    Vios iDx 255, Vios iDx CR 255
    467×467×417 mm
    732×831×110 mm
    1
    1 * 1 ST
    1 * 1 ST - 2 790,00 €
    2 790,00 €
    1 * 1 ST
       2 790,00 €
    Specifications
    Environmentally Preferable
    Capacity
    85 L
    Temperature range
    –50...–86 °C
Shelving System for Nunc Cell Factory System with 10 or 13 layers
  •  
    For
    W×D×H int.
    W×D×H ext.
    Pk
  • Set of 3 shelves to support up to six 10- or 13-layer culture vessels, such as Nunc Cell Factory Systems
    Set of 3 shelves to support up to six 10- or 13-layer culture vessels, such as Nunc Cell Factory Systems
    390-1694
    50166285
    Vios iDx 255, Vios iDx CR 255
    — mm
    — mm
    1
    1 * 1 ST
    1 * 1 ST - 1 940,00 €
    1 940,00 €
    1 * 1 ST
       1 940,00 €
    Specifications
    Environmentally Preferable
    Capacity
    Temperature range
Shelving System for G-Rex Bioreactors
  •  
    For
    W×D×H int.
    W×D×H ext.
    Pk
  • Set of 3 shelves to support up to ten G-Rex 500M-CS bioreactors
    Set of 3 shelves to support up to ten G-Rex 500M-CS bioreactors
    390-1671
    50164781
    Vios iDx 255, Vios iDx CR 255
    — mm
    — mm
    1
    1 * 1 ST
    1 * 1 ST - 3 540,00 €
    3 540,00 €
    1 * 1 ST
       3 540,00 €
    Specifications
    Environmentally Preferable
    Capacity
    Temperature range