Netherlands
Order Entry
Culture plates, Corning® Primaria™
Culture plates, Corning® Primaria™
Supplier:  Corning
Certificates
undefined
Culture plates, Corning® Primaria™
Supplier:  Corning
Certificates
Related Information
Product Information
  • Catalog #
    Supplier #
    No. of wells
    Colour
    Culture area
    Sterile
    Packed
    Availability
    Price
  • Corning® Primaria™ TC-treated, PS
    Corning® Primaria™ TC-treated, PS
    734-0077
    353846
    6
    Clear
    9,6 cm²
    +
    1/tray
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 212,87 €
    212,87 €
    1 * 50 ST
       212,87 €
    Supplier Article Number: 353846
    Local Article Number: BDAA353846
    Specifications:
    Specifications
    Environmentally Preferable
  • Corning® Primaria™ TC-treated, PS
    Corning® Primaria™ TC-treated, PS
    734-0078
    353847
    24
    Clear
    2,0 cm²
    +
    1/tray
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 222,51 €
    222,51 €
    1 * 50 ST
       222,51 €
    Supplier Article Number: 353847
    Local Article Number: BDAA353847
    Specifications:
    Specifications
    Environmentally Preferable