Netherlands
Order Entry
Cole-Parmer® Stuart CC-200 Digital Colony Counters
Cole-Parmer® Stuart CC-200 Digital Colony Counters
Supplier:  Cole Parmer
Certificates
undefined
Cole-Parmer® Stuart CC-200 Digital Colony Counters
Supplier:  Cole Parmer
Specifications
  • Catalog No:
  • STUA14212-06
  • STUA14212-04
  • Electrical:
  • 90 - 230 VAC, 50/60 Hz
  • 90 - 230 VAC, 50/60 Hz
  • W×D×H:
  • 300×310×140 mm
  • 300×310×140 mm
Specifications
Related Information
Product Information
  • Catalog #
    Supplier #
    Model
    Pk
    Availability
    Price
  • Digital colony counter
    Digital colony counter
    STUA14212-04
    14212-04
    CC-200
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 290,00 €
    1 290,00 €
    1 * 1 ST
       1 290,00 €
    Supplier Article Number: 14212-04
    Local Article Number: STUA14212-04
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Electrical
    90 - 230 VAC, 50/60 Hz
    W×D×H
    300×310×140 mm
  • Digital colony counter with IQ/OQ documents
    Digital colony counter with IQ/OQ documents
    STUA14212-06
    14212-06
    CC-200
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 240,00 €
    2 240,00 €
    1 * 1 ST
       2 240,00 €
    Supplier Article Number: 14212-06
    Local Article Number: STUA14212-06
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Electrical
    90 - 230 VAC, 50/60 Hz
    W×D×H
    300×310×140 mm