Netherlands
Order Entry
Powder Spatulas
Powder Spatulas
Certificates
undefined
Powder Spatulas
Product Information
  • Catalog #
    Supplier #
    Length
    Pk
    Availability
    Price
  • Powder spatula, 9 g weight
    Powder spatula, 9 g weight
    USBE3283
    3283
    185 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,20 €
    3,20 €
    1 * 1 ST
       3,20 €
    Supplier Article Number: 3283
    Local Article Number: USBE3283
    Specifications:
    Specifications
    Environmentally Preferable
  • Powder spatula, 10 g weight
    Powder spatula, 10 g weight
    USBE3284
    3284
    210 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,40 €
    3,40 €
    1 * 1 ST
       3,40 €
    Supplier Article Number: 3284
    Local Article Number: USBE3284
    Specifications:
    Specifications
    Environmentally Preferable