Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Cell culture flasks and dishes, fibronectin-coated, CELLCOAT®
Leverancier: Greiner bio-one
Cell culture flasks and petri dishes coated with fibronectin, a high molecular weight glycoprotein found in the extracellular matrix (ECM) and plasma. Spread thinly across the surface of the culture, fibronectin is a substrate that encourages the adherence, proliferation and growth of various cell types. Fibronectin-coated surfaces are used for applications such as the inductive adhesion, proliferation and growth of endothelial cells, fibroblasts, smooth muscle cells, neurons and epithelial cells.
  • Shelf life: 6 months at 2 to 8 °C
  • Other fibronectin-coated culture supports are available on request
login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat
Order
Expand All / Collapse All
Inhoud Omschrijving Cultuuroppervlakte Aanbevolen werkvolume VPE VWR catalogus nummer Hoeveelheid
50 ml Cultuurkolf, met fibronectinecoating 25 cm² 5 - 10 ml 10 392-0257
250 ml Celcultuurkolf, met fibronectinecoating 75 cm² 15 - 38 ml 50 392-0233
650 ml Celcultuurkolf, met fibronectinecoating 175 cm² 20 - 85 ml 50 392-0239
17 ml Celcultuurschaaltje, Ø 60 mm×15 mm, met fibronectinecoating 21 cm² 6 - 7 ml 50 392-0219
70 ml Celcultuurschaaltje, Ø 100 mm×20 mm, met fibronectinecoating 58 cm² 16 - 17 ml 50 392-0244
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid