Netherlands
Order Entry
VWR® ARISTAR®, Tantalum Standard Solution, 10000 mg/L Ta in Water with Hydrofluoric Acid (max. 1%) (from Ta), Standard for ICP
VWR® ARISTAR®, Tantalum Standard Solution, 10000 mg/L Ta in Water with Hydrofluoric Acid (max. 1%) (from Ta), Standard for ICP
Supplier:  VWR Chemicals
Certificates
undefined
VWR® ARISTAR®, Tantalum Standard Solution, 10000 mg/L Ta in Water with Hydrofluoric Acid (max. 1%) (from Ta), Standard for ICP
Supplier:  VWR Chemicals
Certificates
Related Information
Product Information
  • Catalog #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • 456114N
    Tantalum
    Ta
    10000 mg/L
    H₂O/tr HF
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 609,00 €
    609,00 €
    1 * 500 mL
       609,00 €
  • Tantalum
    Ta
    10000 mg/L
    H₂O/tr HF
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 240,00 €
    240,00 €
    1 * 100 mL
       240,00 €
Frequently Bought Together