Ethylbroomacetaat 98%
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
ACRO158590025EA 325 EUR
ACRO158590025 ACRO158591000 ACRO158595000
Ethylbroomacetaat 98%
Ethylbroomacetaat
Formule: BrCH₂COOCH₂CH₃
MW: 167 g/mol
Kookpunt: 159 °C (760 mmHg)
Smeltpunt: –38 °C
Dichtheid: 1,506 g/cm³
Vlampunt: 47 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000191
CAS nummer: 105-36-2
EINECS: 203-290-9
UN: 1603
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4500 to 1.4520 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤300 APHA
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.002 %
Insoluble matter ≤0.01 %