Cyanoacetic acid, Sigma-Aldrich®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
M-Clarity™ quality level = MQ200
8.00252.9034
8.00252.9999
Cyanoacetic acid, Sigma-Aldrich®
Cyanoacetic acid
Formule:
C₃H₃NO₂ MW: 85,06 g/mol Kookpunt: 319 °C (1013 hPa) Smeltpunt: 66 °C Dichtheid: >1,1 g/cm³ (20 °C) Vlampunt: >165 °C |
MDL Nummer:
MFCD00002677 CAS nummer: 372-09-8 EINECS: 206-743-9 ADR: 8,II Merck Index: 13,02718 |
Specification Test Results
Solubility | 1000 g/l (20 °C) |
Melting Point | 65 - 68 °C |
Molar Mass | 85.06 g/mol |
Bulk Density | 700 kg/m3 |
Boiling Point | 108 °C (20 hPa) |
Vapor Pressure | 0.1 hPa (100 °C) |
Flash Point | 107 °C |
pH | 1.6 (50 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...