N-Ethylmaleimide (NEM)

Leverancier: Uptima
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

UPTI7899595EA 0 EUR
UPTI7899595
N-Ethylmaleimide (NEM)
N-Ethylmaleimide (NEM)
Formule: C₆H₇NO₂
MW: 125,13 g/mol
Kookpunt: 210 °C (1013 hPa)
Smeltpunt: 46 °C
MDL Nummer: MFCD00005509
CAS nummer: 128-53-0
EINECS: 204-892-4
UN: 2928
ADR: 6.1,II
Merck Index: 13,03855

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR