Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?

Ammoniak-oplossing 30-33% NH₃ in water ≥99%, Puriss., Fluka™

Leverancier: Honeywell Chemicals
Synonyms: AmmoniumhydroxideWaterig-ammoniakale oplossing

Formule: NH₃
MW: 35,05 g/mol
Kookpunt: 32 - 37,7 °C (1013 hPa)
Smeltpunt: -60 - -57,5 °C
Dichtheid: 0,88 to 0.91 g/cm³ (20 °C)
Storage Temperature: Kamertemperatuur
MDL Nummer: MFCD00066650
CAS nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672

login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat


Expand All / Collapse All
Pack type VPE VWR catalogus nummer Hoeveelheid
Glass Bottle 6 l 05002-6X1L
Glass Bottle 1 KIT 05002-4X2.5L
Glass Bottle, Narrow Open 1 l 05002-1L
Glass Bottle, Narrow Open 2,5 l 05002-2.5L
Paper / Plastic Bag 22 kg 05002-22KG
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid