Hydrazine hydrate 4.8% in aqueous solution, VOLUSOL® volumetric solution
VWRS83915.560
:
- Pack type:Plastic drum
- Pk:200 L
- Seq:5549
- Assay (calculated as N2H4.H2O):4.7 4.9 %
- pH:9 11
- Cl (Chloride):Max. 0.5 ppm
- F (Fluoride):Max. 0.5 ppm
- Si (Silicium):Max. 0.5 ppm
- SO4 (Sulphate):Max. 1 ppm
- Solids in suspension:Max. 0.1 ppm
:
:32.05 g/mol (anhy)
:∼100 °C (1013 hPa)
:∼1.02 g/cm³ (20 °C)
:206-114-9
:3293
:6.1,III
:GHS06
:GHS08
:GHS09
:10217-52-4
:7803-57-8
:NH₂NH₂·xH₂O
:MFCD00149931