Netherlands
Order Entry
VWR® Mega Racks® Tube Racks for 28 to 32 mm Tubes
VWR® Mega Racks® Tube Racks for 28 to 32 mm Tubes
Supplier:  VWR Collection
New Product
Certificates
undefined
VWR® Mega Racks® Tube Racks for 28 to 32 mm Tubes
Supplier:  VWR Collection
New Product
Product Information
  • Catalog #
    Hole Ø
    Colour
    No. of Holes
    For tubes
    W×D×H
    Pk
    Availability
    Price
  • 525-1619
    32 mm
    White
    100
    50 ml
    412×423×120 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,90 €
    64,90 €
    1 * 1 ST
       64,90 €
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • 525-1620
    32 mm
    Blue
    100
    50 ml
    412×423×120 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,90 €
    64,90 €
    1 * 1 ST
       64,90 €
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product