Netherlands
Order Entry
Locator™ 6 Rack and Box System
Locator™ 6 Rack and Box System
Certificates
undefined
Locator™ 6 Rack and Box System
Specifications
  • Catalog No:
  • BARNCY509113
  • 478-0808
  • Capacity:
  • 184 L
  • 184 L
  • Ø×H:
  • 953×660 mm
  • 953×660 mm
Specifications
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Locator 6 with level monitor, manual fill LN₂ dewar, 110 - 220 V 50 Hz
    Locator 6 with level monitor, manual fill LN₂ dewar, 110 - 220 V 50 Hz
    BARNCY509113
    CY509113
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Supplier Article Number: CY509113
    Local Article Number: BARNCY509113
    Specifications:
    More
    Specifications
    Capacity
    184 L
    Environmentally Preferable
    Ø×H
    953×660 mm
  • Locator 6 without level monitor, manual fill LN₂ dewar, 110 - 220 V 50 Hz
    Locator 6 without level monitor, manual fill LN₂ dewar, 110 - 220 V 50 Hz
    478-0808
    CY50985
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 650,00 €
    7 650,00 €
    1 * 1 ST
       7 650,00 €
    Supplier Article Number: CY50985
    Local Article Number: BARNCY50985
    Specifications:
    More
    Specifications
    Capacity
    184 L
    Environmentally Preferable
    Ø×H
    953×660 mm