Starch potassium iodide paper Reag.Ph.Eur., Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.09512.0003EA 26.25 EUR
1.09512.0003
Starch potassium iodide paper Reag.Ph.Eur., Supelco®
Testpapieren Indicatorpapier
Potassium iodine paper is used for the determination of oxidising agents.